Skip to Content
Merck
All Photos(2)

Key Documents

386A

Sigma-Aldrich

Leukocyte Acid Phosphatase (TRAP) Kit

Select reagents packaged in gelatin capsules

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.47

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Principle

Peripheral blood or bone marrow preparations are fixed to a microscope slide. The resulting film is incubated in a solution of Naphthol AS-BI phosphoric acid and Fast Garnet GBC. Kit is used to demonstrate acid phosphatase and tartrate resistant acid phosphatase (TRAP) in blood, bone marrow and tissue touch preparations.

Kit Components Only

Product No.
Description

  • Acetate Solution (386-3) 50 mL

  • Acid Hematoxylin (285-2) 100 mL

  • Citrate Concentrate (386-1) 20 mL

  • Fast Garnet GBC Salt (kit only) 2 × 10

  • Naphthol AS-BI Phosphoric Acid (386-4) 40 mL

  • Tartrate Solution (kit only) 20 mL

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annina Sipola et al.
BMC musculoskeletal disorders, 7, 56-56 (2006-07-15)
Endostatin is a C-terminal fragment of collagen XVIII which is a component of basement membranes with the structural properties of both collagens and proteoglycans. Endostatin has a major role in angiogenesis which is intimately associated with bone development and remodeling.
Gui-Hua Quan et al.
International journal of molecular sciences, 16(12), 29496-29507 (2015-12-23)
Calycosin, an isoflavonoid phytoestrogen, isolated from Radix Astragali, was reported to possess anti-tumor, anti-inflammation, and osteogenic properties, but its impact on osteoclast differentiation remains unclear. In this study, we examined the effects of calycosin on osteoclastogenesis induced by RANKL. The
Yuri Akita et al.
Journal of bone and mineral metabolism, 36(5), 547-559 (2017-10-19)
Osteonecrosis of the jaw (ONJ), which is a rare but severe adverse effect, mainly occurs in oncology patients receiving chemotherapeutic agents and bisphosphonates. However, the combined impact of chemotherapy and bisphosphonates on wound healing after tooth extraction remains unknown. The
Zhenheng Wang et al.
International journal of nanomedicine, 12, 5387-5397 (2017-08-11)
Wear particle-induced inflammatory osteolysis is the primary cause of aseptic loosening, which is the most common reason for total hip arthroplasty (THA) failure in the med- and long term. Recent studies have suggested an important role of gut microbiota (GM)
Yen-You Lin et al.
Journal of advanced research, 24, 109-120 (2020-04-08)
Deposition of monosodium urate (MSU) crystals in the joint or synovium is the major factor in Gouty arthritis (GA). The clinical features of chronic and recurrent GA include pain and the subsequent development of chronic tophaceous GA with multiple tophi

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service