Skip to Content
Merck
All Photos(1)

Key Documents

38132

Sigma-Aldrich

N-(5-Fluoresceinyl)maleimide

≥90% (HPLC), BioReagent, suitable for fluorescence

Synonym(s):

5-Maleimido-fluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H13NO7
CAS Number:
Molecular Weight:
427.36
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

≥90% (HPLC)

fluorescence

λex 490 nm; λem 518 nm in 0.1 M Tris pH 8.0

suitability

corresponds for coupling to thiols
suitable for fluorescence

storage temp.

2-8°C

SMILES string

Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5cc(ccc45)N6C(=O)C=CC6=O)c1

InChI

1S/C24H13NO7/c26-13-2-5-17-19(10-13)31-20-11-14(27)3-6-18(20)24(17)16-4-1-12(9-15(16)23(30)32-24)25-21(28)7-8-22(25)29/h1-11,26-27H

InChI key

AYDAHOIUHVUJHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-(5-Fluoresceinyl) maleimide, also known as fluorescein-5-maleimide (FM), is a fluorescent derivatization reagent. The pH during the derivatization reaction plays a critical role.

Application

N-(5-Fluoresceinyl) maleimide or fluorescein-5-maleimide is used as a fluorescent biosensor to detect nanoparticles.

N-(5-Fluoresceinyl) maleimide (5-FM) is used to fluorescence label molecules such as proteins and peptides via their thiol groups.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Giangregorio et al.
Biochimica et biophysica acta, 1767(11), 1331-1339 (2007-10-27)
During substrate translocation mitochondrial carriers cycle between the cytoplasmic-state (c-state) with substrate-binding site open to the intermembrane space and matrix-state (m-state) with the binding site open to the mitochondrial matrix. Here, the accessibility of Cys-58, Cys-136 and Cys-155 of the
Oluyemi Akinloye et al.
Analytical and bioanalytical chemistry, 409(4), 903-915 (2016-12-03)
The ability to detect and quantify nanoparticles is essential but there is currently no simple, sensitive, and rapid method for the detection of nanomaterials. We have developed a novel peptide-based fluorescence-based biosensor for detection and measurement of negatively charged engineered
Robert E Dempski et al.
The Journal of biological chemistry, 281(47), 36338-36346 (2006-09-19)
The Na+/K+-ATPase maintains the physiological Na+ and K+ gradients across the plasma membrane in most animal cells. The functional unit of the ion pump is comprised of two mandatory subunits including the alpha-subunit, which mediates ATP hydrolysis and ion translocation
Di Zhou et al.
Macromolecular bioscience, 12(4), 465-474 (2012-04-13)
Branched poly(methoxy-PEG acrylate) and thermally responsive poly(methoxy-PEG acrylate)-block-poly(N-isopropylacrylamide) are synthesized by RAFT polymerization. After reduction, these polymers are fluorescently labeled by reacting the free thiol groups with N-(5-fluoresceinyl)maleimide. As shown by DLS, the labeled copolymer poly(methoxy-PEG acrylate)-block-poly(N-isopropylacrylamide) forms nanoparticles at
Michal Sharoni et al.
The Journal of biological chemistry, 280(38), 32849-32855 (2005-07-29)
EmrE is a small multidrug transporter in Escherichia coli that extrudes various positively charged drugs across the plasma membrane in exchange with protons, thereby rendering cells resistant to these compounds. Biochemical experiments indicate that the basic functional unit of EmrE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service