Skip to Content
Merck
All Photos(2)

Key Documents

360376

Sigma-Aldrich

Capsaicin

63.7% (HPLC), powder, TRPV1 agonist

Synonym(s):

8-Methyl-N-vanillyl-trans-6-nonenamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH=CH(CH2)4CONHCH2C6H3-4-(OH)-3-(OCH3)
CAS Number:
Molecular Weight:
305.41
Beilstein:
2816484
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Capsaicin, natural

grade

natural

Assay

63.7% (HPLC)

form

powder

composition

capsaicin, 65%
dihydrocapsaicin, 35%

mp

62-65 °C (lit.)

solubility

chloroform: 5%

storage temp.

2-8°C

SMILES string

COc1cc(CNC(=O)CCCC\C=C\C(C)C)ccc1O

InChI

1S/C18H27NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h6,8,10-12,14,20H,4-5,7,9,13H2,1-3H3,(H,19,21)/b8-6+

InChI key

YKPUWZUDDOIDPM-SOFGYWHQSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Capsaicin (Zostrix), an alkaloid extract is obtained from hot chili peppers. It is accessible in the form of cream and lotion. Capsaicin is a hydrophobic, instable and pungent white crystalline powder, that has no smell and color. It is produced by mixing a branched-chain fatty acid to vanillylamine in the chili pepper.

Application

Capsaicin has been used in the development of the isolation methods. It has also been used in the development of enzyme based electrochemical biosensor for the determination of capsaicin using a novel signal amplification strategy.

Biochem/physiol Actions

Capsaicin is used as an important component of oleoresin capsicum or pepper spray. It plays a beneficial role in obesity, cardiovascular and gastrointestinal conditions, several cancers, neurogenic bladder and dermatologic conditions. It is used in the production of spices, chilli sauces and pain-inducing defensive pepper sprays. Capsaicin is also used to manufacture creams to treat diabetic neuropathy, cluster headache, psoriasis, rheumatoid arthritis and reflex sympathetic dystrophy. It is considered as an agonist of the transient receptor potential cation channel subfamily V member 1 (TrpV1).
Prototype vanilloid receptor agonist. Neurotoxin; activates sensory neurons that give rise to unmyelinated C-fibers, many of which contain substance P. Topical application desensitizes the sensory nerve endings giving a paradoxical antinociceptive effect; systemic administration can be neurotoxic to capsaicin-sensitive cells, especially in newborn animals. Active component of chili peppers.
Prototype vanilloid receptor agonist; neurotoxin. Active component of chili peppers.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanisms and clinical uses of capsaicin
Sharma S K, et al.
European Journal of Pharmacology, 720(1-3), 55-62 (2013)
Isolation of individual capsaicinoids from a mixture and their characterization by 13C NMR spectrometry
Thompson R Q, et al.
Talanta, 70(2), 315-322 (2006)
Electrochemical sensing platform amplified with a nanobiocomposite of L-phenylalanine ammonia-lyase enzyme for the detection of capsaicin
Sabela M I, et al.
Biosensors And Bioelectronics, 45-53 (2016)
Postherpetic neuralgia
Christo P J and Cauley B D
Headache, 48, 130-134 (2008)
Capsaicin, the primary constituent of pepper sprays and its pharmacological effects on mammalian ocular tissues
Krishnatreyya H, et al.
European Journal of Pharmacology, 819, 114-121 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service