Skip to Content
Merck
All Photos(3)

Key Documents

31410

Sigma-Aldrich

Dextrin from maize starch

Synonym(s):

Maltodextrin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

maize

description

dextrose equivalent 8.0 - 15.0

form

powder

impurities

≤0.5% sulfated ash

color

white

mp

240  °C ((464 °F ))

storage temp.

room temp

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6+/m0/s1

InChI key

GZCGUPFRVQAUEE-SLPGGIOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Maltodextrin, a polymer chain composed of D-glucose units linked by α (1,4) bonds, is used in the development of encapsulation and nanencapsulation vehicles for use in the food industry. Maltodextrin is used as a protein Maillard-induced conjugation agent for improved solubility and foaming activity. As a sugar-matrix forming agent, maltodextrin is used in the development of orally disintegrating tablets (ODT).

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gilad Markman et al.
Food & function, 3(3), 262-270 (2011-12-24)
Maillard reaction based protein-polysaccharide conjugates (PPC) are typically better emulsifiers than the proteins used to form them. Such conjugates have been proposed as encapsulation agents, but their potential as nanoencapsulation agents in clear beverages has not yet been proposed. In
N V Grigorovich et al.
Food & function, 3(3), 283-289 (2011-12-14)
The sodium caseinate (SCN)-maltodextrin (MD) covalent conjugates were prepared by a food-grade process involving the first step of the Maillard reaction. The covalent conjugates were prepared with different weight ratios of biopolymers (R(MD : SCN) = 0.4; 1; 2; 5) in the
Development and optimization of lyophilized orally disintegrating tablets using factorial design.
Ahmed IS, Shamma RN, Shoukri RA.
Pharmaceutical Development and Technology, doi: 10-doi: 10 (2011)
Yosra Shaaban R Elnaggar et al.
AAPS PharmSciTech, 11(2), 645-651 (2010-04-21)
The recent challenge in orally disintegrating tablets (ODT) manufacturing encompasses the compromise between instantaneous disintegration, sufficient hardness, and standard processing equipment. The current investigation constitutes one attempt to fulfill this challenge. Maltodextrin, in the present work, was utilized as a
Evaluation of rapidly disintegrating tablets manufactured by phase transition of sugar alcohols.
Kuno Y, Kojima M, Ando S, Nakagami H.
J. Controlled Release, 105, 16-22 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service