Skip to Content
Merck
All Photos(1)

Key Documents

31170

Sigma-Aldrich

2-Deoxy-D-ribose

≥99.0% (TLC)

Synonym(s):

2-Deoxy-D-arabinose, 2-Deoxy-D-erythropentose, Thyminose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O4
CAS Number:
Molecular Weight:
134.13
Beilstein:
1721978
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99.0% (TLC)

form

solid

optical activity

[α]20/D −56±2°, 24 hr, c = 1% in H2O

impurities

<0.5% Sulphated ash

ign. residue

≤0.5% (as SO4)

loss

≤1% loss on drying, 20 °C (HV)

color

white

mp

89-90 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)CC=O

InChI

1S/C5H10O4/c6-2-1-4(8)5(9)3-7/h2,4-5,7-9H,1,3H2/t4-,5+/m0/s1

InChI key

ASJSAQIRZKANQN-CRCLSJGQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Deoxy-D-ribose is used to study processes of oxidative stress and glycation in vivo and in vitro. 2-Deoxy-D-ribose, an endothelial-cell chemoattractant and angiogenesis-inducing factor, is used to study processes of tumor angiogenesis and progression mediated at the level of thymidine phosphorylase activity.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tymofii Yu Nikolaienko et al.
Physical chemistry chemical physics : PCCP, 14(44), 15554-15561 (2012-10-18)
Relaxed force constants (RFC) and vibrational root-mean-square (VRMS) deviations are used for comparative characterization of mechanical properties of canonical 2'-deoxyribonucleosides (2DRs) and 1,2-dideoxyribose molecule, their model sugar residue. It has been shown that RFC and VRMS should be preferred over
Manuel Ellermann et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 3), 253-260 (2012-02-22)
The biological activity of catechol neurotransmitters such as dopamine in the synapse is modulated by transporters and enzymes. Catechol-O-methyltransferase (COMT; EC 2.1.1.6) inactivates neurotransmitters by catalyzing the transfer of a methyl group from S-adenosylmethionine to catechols in the presence of
Buthina Al-Oudat et al.
Bioorganic & medicinal chemistry letters, 23(3), 854-859 (2012-12-26)
Well-defined substrates for the study of oxidative processes are important for the elucidation of the role of DNA damage in the etiology of diseases such as cancer. We have synthesized 3'-modified oligodeoxyribonucleotides (ODNs) using 5'→3' 'reverse' DNA synthesis for the
Anita R Peoples et al.
Nucleic acids research, 40(13), 6060-6069 (2012-04-03)
Our mechanistic understanding of damage formation in DNA by the direct effect relies heavily on what is known of free radical intermediates studied by EPR spectroscopy. Bridging this information to stable product formation requires methods with comparable sensitivities, a criterion
Adam J Stevens et al.
Biochemistry, 52(5), 975-983 (2013-01-22)
Base excision repair (BER) plays a vital role in maintaining genomic integrity in mammalian cells. DNA polymerase λ (Pol λ) is believed to play a backup role to DNA polymerase β (Pol β) in base excision repair. Two oxidized abasic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service