Skip to Content
Merck
All Photos(1)

Documents

21878

Sigma-Aldrich

5(6)-Carboxyfluorescein N-hydroxysuccinimide ester

BioReagent, suitable for fluorescence, mixture of isomers, ≥80% (HPLC)

Synonym(s):

5(6)-FAM SE, FLUOS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H15NO9
CAS Number:
Molecular Weight:
473.39
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥80% (HPLC)

form

powder

solubility

DMF: soluble
DMSO: soluble

fluorescence

λex 492 nm; λem 517 nm in DMF

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

OC(=O)c1ccc(cc1C2=C3C=CC(=O)C=C3Oc4cc(O)ccc24)C(=O)ON5C(=O)CCC5=O.OC(=O)c6cccc(C(=O)ON7C(=O)CCC7=O)c6C8=C9C=CC(=O)C=C9Oc%10cc(O)ccc8%10

InChI

1S/2C25H15NO9/c27-13-2-5-16-19(10-13)34-20-11-14(28)3-6-17(20)23(16)18-9-12(1-4-15(18)24(31)32)25(33)35-26-21(29)7-8-22(26)30;27-12-4-6-14-18(10-12)34-19-11-13(28)5-7-15(19)22(14)23-16(24(31)32)2-1-3-17(23)25(33)35-26-20(29)8-9-21(26)30/h1-6,9-11,27H,7-8H2,(H,31,32);1-7,10-11,27H,8-9H2,(H,31,32)

InChI key

YVTPVJDECOUTJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5(6)-FAM SE, FLUOS) may be used to prepare fluorescent derivatives of proteins such as tubulin and for the preparation of a phenytoin tracer for the determination of this and other drugs in serum and plasma by fluorescence polarization immunoassay (FPIA) and laser-induced fluorescence detection.
Fluorescent label for primary and secondary amine groups on proteins and other biomolecules.
Has been used to prepare fluorescent derivatives of tubulin and for the preparation of a phenytoin tracer for the determination of this drug in serum and plasma by fluorescence polarization immunoassay (FPIA)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miki Arai Hojo et al.
Nature communications, 10(1), 2603-2603 (2019-06-15)
During thymic negative selection, autoreactive thymocytes carrying T cell receptor (TCR) with overtly strong affinity to self-MHC/self-peptide are removed by Bim-dependent apoptosis, but how Bim is specifically regulated to link TCR activation and apoptosis induction is unclear. Here we identify
Mingze Chang et al.
Oncology reports, 38(2), 1251-1258 (2017-07-06)
microRNA-33a (miR-33a) belongs to the miR-33 family that is implicated in the progression of various types of cancers. Aberrant expression of miR-33a has been detected in several human cancers, and has been shown to regulate the migration and invasion as
Hsin-Hua Yeh et al.
Electrophoresis, 31(11), 1903-1911 (2010-05-07)
A nonionic surfactant MEKC method with LIF detection was developed for the simultaneous determination of memantine, an anti-Alzheimer's disease agent, and amantadine, an anti-Parkinson's disease drug, in human plasma. Before analysis, the plasma samples were pretreated by liquid-liquid extraction with
Alma Mariana Fuentes-González et al.
Archives of virology, 164(7), 1815-1827 (2019-01-12)
Persistent infections with high-risk human papillomaviruses (HR-HPVs) are linked to the development of cervical cancer due to a deregulation of the productive viral cycle in the host cell, leading to cell transformation. The E2 viral protein is expressed early during an
Teruhisa Takagishi et al.
Scientific reports, 7(1), 5217-5217 (2017-07-14)
Clostridium perfringens α-toxin induces hemolysis of erythrocytes from various species, but it has not been elucidated whether the toxin affects erythropoiesis. In this study, we treated bone marrow cells (BMCs) from mice with purified α-toxin and found that TER119

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service