Skip to Content
Merck
All Photos(1)

Key Documents

21056

Sigma-Aldrich

Calcium acetate hydrate

BioUltra, ≥99.0% (calc. on dried material, KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Ca · xH2O
CAS Number:
Molecular Weight:
158.17 (anhydrous basis)
Beilstein:
3692527
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.26

product line

BioUltra

Assay

≥99.0% (calc. on dried material, KT)

form

powder

impurities

insoluble matter, passes filter test
≤7% water (Karl Fischer)

pH

7.0-8.5 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

As: ≤0.1 mg/kg
Ba: ≤20 mg/kg
Bi: ≤5 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤100 mg/kg
Li: ≤5 mg/kg
Mg: ≤1000 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤150 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤200 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.090
λ: 280 nm Amax: 0.070

SMILES string

O.CC(=O)O[Ca]OC(C)=O

InChI

1S/2C2H4O2.Ca.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

XQKKWWCELHKGKB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Adsorption of Lead Ion from Wastewater Using Non-Crystal Hydrated Calcium Silicate Gel:</strong> This study demonstrates the utility of calcium acetate hydrate in the synthesis of calcium silicate gel for environmental applications, specifically for the adsorption of heavy metals from wastewater, aligning with its use in water treatment processes (Liu et al., 2021).</li>
<li><strong>Dexamethasone Loaded Liposomes by Thin-Film Hydration and Microfluidic Procedures: Formulation Challenges:</strong> Research explores the use of calcium acetate hydrate in pharmaceutical formulations, particularly in the stabilization of liposome structures, which are critical in drug delivery systems (Al-Amin et al., 2020).</li>
<li><strong>Liposomal diclofenac eye drop formulations targeting the retina: formulation stability improvement using surface modification of liposomes:</strong> Discusses the enhancement of ocular drug delivery systems using calcium acetate hydrate, focusing on its role in improving the stability and bioavailability of therapeutic agents (Fujisawa et al., 2012).</li>
</ul>

Other Notes

Isolation of plasma membranes from HeLa cells; Salt of choice in aequorin bioluminescence assay; Protein precipitant in the manufacture of sunflower protein; Alleviates defoliation and fruit drop induced by Ethephon in Citrus reticulata

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E.M. Bau et al.
Bull. Liaison Groupe Polyphenols, 8, 36-36 (1978)
S. Iwahori et al.
Sci. Hortic., 12, 265-265 (1980)
E.H. Eylar et al.
Methods in Enzymology, 22, 124-124 (1971)
F.H. Johnson et al.
Methods in Enzymology, 57, 278-278 (1978)
Yang Xu et al.
Autophagy, 15(6), 1017-1030 (2019-01-18)
As a master regulator of the macroautophagy/autophagy-lysosomal pathway, TFEB (transcription factor EB) plays a prominent role in regulating neurodegenerative diseases and cancer. The transcription activity of TFEB is tightly controlled by phosphorylation and dephosphorylation. Phosphorylated S211 (p-S211) of TFEB can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service