Skip to Content
Merck
All Photos(1)

Key Documents

15S7

Sigma-Aldrich

TERGITOL

greener alternative

Type 15-S-7

Synonym(s):

Secondary alcohol ethoxylate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161900
NACRES:
NA.25

description

non-ionic

Quality Level

type

Type 15-S-7

form

liquid

mol wt

508 g/mol

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

CMC

38 ppm

transition temp

pour point 1 °C
cloud point 37 °C

HLB

12.1

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

Tergitol 15-S-7 is a linear non-ionic surfactant that is a secondary ethoxylated alcohol. It is compatible with anionic, cationic, and other nonionic surfactants.

Application

Tergitol 15-S-7 has been used:
  • To study the synergistic effect of Tergitol 15-S-7 and AOT (dioctyl sulfosuccinate) on bacterial oxidation of alkanes in crude oils
  • In cloud-point extraction technique to extract polycyclic aromatic hydrocarbons from aqueous solutions
  • Along with MgSO4 in anolyte reservoir for electrokinetic experiments
  • As a very efficient surfactant in removing non-polar oil
  • To study the phase behavior of surfactant-brine–oil system, which is desired for enhanced oil recovery

Features and Benefits

  • Excellent formulation & handling properties
  • Superior wetting & rinseability
  • Low odor
  • Narrow gel range

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

469.4 °F - open cup

Flash Point(C)

243 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rates of solubilization of triolein into nonionic surfactant solutions.
Chen BH, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 128(1-3), 129-143 (1997)
Electrokinetic remediation of contaminated soil with waste-lubricant oils and zinc.
Park SW, et al.
Journal of Hazardous Materials, 169(1-3), 1168-1172 (2009)
Behavior of surfactant mixtures in model oily-soil detergency studies.
Aronson M P, et al.
Journal of the American Oil Chemists' Society, 60(7), 1333-1339 (1983)
Effects of additives on the cloud points of selected nonionic linear ethoxylated alcohol surfactants.
Li J L, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 346(1-3), 237-243 (2009)
Determination of estrogens in human urine by high-performance liquid chromatography/diode array detection with ultrasound-assisted cloud-point extraction.
Zou Y, et al.
Analytical Biochemistry, 421(2), 378-384 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service