Skip to Content
Merck
All Photos(1)

Documents

147M-9

Sigma-Aldrich

CD45RO (UCHL-1) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

UCHL-1, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (147M-94)
vial of 0.5 mL concentrate (147M-95)
bottle of 1.0 mL predilute (147M-97)
vial of 1.0 mL concentrate (147M-96)
bottle of 7.0 mL predilute (147M-98)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG2aκ

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... PTPRC(5788)

General description

Anti-CD45RO (T-Cell, Pan) antibody reacts with thymocytes and activated T-cells, but only on a subpopulation of resting T-cells. This antibody shows no reactivity with B-cells making it a good marker for T-cell tumors. In addition, granulocytes and monocytes are also labeled with this antibody. T-Cell, Pan has been designated as CD45RO at The International Leukocyte Typing Workshop.

Quality


IVD

IVD

IVD

RUO

Linkage

CD45RO Positive Control Slides, Product No. 147S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Falini et al.
Human pathology, 21(6), 624-629 (1990-06-01)
Monoclonal antibodies (mAbs) directed against the leucocyte common (CD45) antigen have been proposed as a useful tool for the differential diagnosis between malignant lymphomas (CD45+) and poorly differentiated nonhemopoietic tumors (CD45-). Thanks to the availability of mAbs directed against fixative-resistant
J A Tworek et al.
American journal of clinical pathology, 110(5), 582-589 (1998-11-05)
Small lymphocytic lymphoma (SLL), mantle cell lymphoma (MCL), and SLL plasmacytoid (SLLP) are malignant neoplasms of small B cells that may have overlapping cytologic features. Entities such as SLL with irregular nuclear contours may pose additional diagnostic difficulties. We investigated
J H Ritter et al.
Journal of cutaneous pathology, 21(6), 481-493 (1994-12-01)
In the skin, separation of selected lymphomas from lymphoid hyperplasia can be challenging. The authors examined 45 cutaneous lymphomas, excluding mycosis fungoides (26 small lymphocytic or mixed tumors; 19 large cell lymphomas), 10 "atypical" lesions, and 40 lesions of presumed
P A Hall et al.
Journal of clinical pathology, 40(2), 151-156 (1987-02-01)
The use of the murine monoclonal antibody MB2 for identifying B lymphocytes in routinely processed tissue was evaluated and contrasted with the use of the monoclonal antibody UCHL1 for identifying T cells. One hundred and sixty eight surgical biopsy specimens
A E Koch et al.
Journal of clinical immunology, 10(4), 192-199 (1990-07-01)
We have characterized the lymphocytes in the synovium of patients with rheumatoid arthritis (RA) by immunohistochemistry using monoclonal antibodies directed against B lymphocytes, T lymphocytes, and antibodies directed against CD45RA and CD45RO, which define T-cell subsets. Both CD45RA+ and CD45RO+

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service