Skip to Content

Legal compliance reminder: purchasers must obtain all necessary licenses and comply with applicable laws (see General Terms & Conditions of Sale). We reserve the right to request additional information.

Merck

Key Documents

Skip To

14533

bisBenzimide H 33342 trihydrochloride

suitable for fluorescence, ≥97.0% (HPLC)

Synonym(s):

2′-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi-1H-benzimidazole trihydrochloride, HOE 33342, Hoechst 33342, bisBenzimide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C27H28N6O · 3HCl · xH2O
CAS Number:
Molecular Weight:
561.93 (anhydrous basis)
UNSPSC Code:
12161505
NACRES:
NA.25
PubChem Substance ID:
EC Number:
245-691-1
Beilstein/REAXYS Number:
1234011
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (HPLC)

fluorescence

λex 340 nm; λem 510 nm, λex 355 nm; λem 465 nm in TE buffer; DNA

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Cl[H].CCOc1ccc(cc1)C2=NCc3cc(ccc3N2)C4=NCc5cc(ccc5N4)N6CCN(C)CC6

InChI

1S/C29H32N6O.3ClH/c1-3-36-25-8-4-20(5-9-25)28-30-18-22-16-21(6-10-26(22)32-28)29-31-19-23-17-24(7-11-27(23)33-29)35-14-12-34(2)13-15-35;;;/h4-11,16-17H,3,12-15,18-19H2,1-2H3,(H,30,32)(H,31,33);3*1H

InChI key

FYEVKHPLBHLWHK-UHFFFAOYSA-N

Application

Useful for staining DNA, chromosomes and nuclei. May be used for fluorescence microscopy or flow cytometry.
Excitation max. = 346 nm
Emission max. = 460 nm
bisBenzimide H 33342 trihydrochloride can be used to study biochemicals and reagents, fluorescent probes, labels, particles and stains, luminescent compounds and detection and wavelength index. bisBenzimide H 33342 trihydrochloride, Hoechst 33342, has been used in a study to investigate the effects of inorganic and organic arsenic compounds on human T-lymphoblastoid leukemia cells. Hoechst 33342 has also been used in a study to examine the effects of 9 calcium antagonists on ABCG2/BCRP-mediated resistance and transport in HeLa and SN-38-resistant HeLa (HeLa/SN100) cells, overexpressing ABCG2/BCRP.

Biochem/physiol Actions

Membrane-permeable, fluorescent DNA stains with low cytotoxicity that intercalate in A-T regions of DNA.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Suitable for vital DNA staining of a variety of cell types; As a probe of membrane permeability in mammalian cells

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B2261B2883382061
description

suitable for fluorescence, ≥97.0% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

Membrane-permeable, adenine-thymine-specific fluorescent stain.

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

passes application test for fluorescence

suitability

-

fluorescence

λex 340 nm; λem 510 nm, λex 355 nm; λem 465 nm in TE buffer; DNA

fluorescence

-

fluorescence

-

fluorescence

λex 346 nm, λem 460 nm

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

100

assay

≥97.0% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–6 of 6 Questions  
  1. Is it necessary to fix and permeabilize to use hoechst with oocytes?

    1 answer
    1. Hoechst 33342 is commonly used for live-cell imaging. Fixation is not required as this material is more lipophilic as compared to the Hoechst 33258. This product has not been tested specifically on oocytes, however several publication referencing this material were found in a literature search. Please see the link below to review one example:
      Effect of Hoechst 33342 staining on developmental competence of prepubertal goat oocytes
      https://pubmed.ncbi.nlm.nih.gov/12214800/

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. When Product 14533, bisBenzimide H 33342 trihydrochloride, is dissolved in water, what is the pH?

    1 answer
    1. The pH of a 2% solution in water is approximately 1.9.

      Helpful?

  6. What is the solubility of Product 14533, bisBenzimide H 33342 trihydrochloride?

    1 answer
    1. It is soluble in water at 50 mg/mL.  It will precipitate from phosphate buffer solutions

      Helpful?

Reviews

No rating value

Active Filters