Skip to Content
Merck
All Photos(1)

Key Documents

121M-1

Sigma-Aldrich

CD21 (2G9) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

2G9, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (121M-14)
vial of 0.5 mL concentrate (121M-15)
bottle of 1.0 mL predilute (121M-17)
vial of 1.0 mL concentrate (121M-16)
bottle of 7.0 mL predilute (121M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG2a

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... CR2(1380)

General description

CD21 (also known as complement receptor 2 (CR2), C3d receptor, or EBV receptor) is a 140 kDa membrane protein on B-lymphocytes to which the Epstein-Barr virus (EBV) binds during infection of these cells. The antigen is absent on T-lymphocytes, monocytes, and granulocytes.
Anti-CD21 is useful in the identification of follicular dendritic cell matrix found in normal lymph nodes and tonsillar tissue. This antibody also labels follicular dendritic cell sarcomas.

Anti-CD21 is valuable in differentiating follicular lymphoma with marginal zone differentiation from marginal zone lymphoma with follicular involvement. It also plays a role in distinguishing among nodular lymphocyte predominant Hodgkin lymphoma, lymphocyte-rich classic Hodgkin lymphoma, and T-cell/histiocyte-rich B-cell lymphoma in combination with other B-cell and T-cell markers.Anti-CD21 is also useful in identifying abnormal follicular dendritic cell pattern in angioimmunoblastic T-cell lymphoma and follicular T-cell lymphoma.

Linkage

CD21 Positive Control Slides, Product No. 121S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-Sung Chuang et al.
International journal of hematology, 91(4), 687-691 (2010-03-04)
Angioimmunoblastic lymphoma (AITL) is a nodal peripheral T-cell lymphoma characterized by a proliferation of arborizing vessels and hyperplastic follicular dendritic cells as well as a polymorphous lymphoid infiltrate including neoplastic cells with clear cytoplasm. Adult T-cell leukemia/lymphoma (ATLL) is caused
A C Chan et al.
Histopathology, 38(6), 510-518 (2001-06-26)
Hyaline-vascular Castleman's disease (HVCD) and follicular dendritic cell (FDC) sarcoma occurring in the nasopharynx are both extremely rare. We report the first case of transformation of the former into the latter as documented by sequential biopsies. The steps involved in
Kong-Chao Chang et al.
The Journal of pathology, 201(3), 404-412 (2003-11-05)
Follicular dendritic cells (FDCs) in reactive germinal centres (GCs) show modulated expression of antigens indicative of step-wise maturation from more undifferentiated stroma. The present study compared the expression of FDC markers CD21, CD23, CD35, and chemokine CXCL13 and the stromal
W Cheuk et al.
The American journal of surgical pathology, 25(6), 721-731 (2001-06-08)
Follicular dendritic cell (FDC) tumors are uncommon neoplasms that can involve lymph nodes or extranodal sites. They can exhibit a broad spectrum of histologic appearances and behavior, but the intra-abdominal ones usually pursue an aggressive course. The purpose of this
David A Biddle et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 15(1), 50-58 (2002-01-18)
Extranodal follicular dendritic cell (FDC) sarcoma of the head and neck region is uncommon, with 16 well-documented cases previously reported (four in the tonsil, four in the pharynx, two in the palate, five in the soft tissue, and one in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service