Skip to Content
Merck
All Photos(1)

Key Documents

08445

Sigma-Aldrich

7-Amino-4-methyl-3-coumarinylacetic acid

BioReagent, suitable for fluorescence, ~90% (HPLC)

Synonym(s):

2H-1-Benzopyran-3-acetic acid, AMCA-H, Aminomethyl coumarin acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO4
CAS Number:
Molecular Weight:
233.22
Beilstein:
7927205
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

Assay

~90% (HPLC)

form

powder

solubility

DMF: soluble
DMSO: soluble
aqueous base: soluble

fluorescence

λex 350 nm; λem 433 nm in methanol

suitability

suitable for fluorescence

SMILES string

CC1=C(CC(O)=O)C(=O)Oc2cc(N)ccc12
CC1=C(CC(O)=O)C(=O)Oc2cc(N)ccc12

InChI

1S/C12H11NO4/c1-6-8-3-2-7(13)4-10(8)17-12(16)9(6)5-11(14)15/h2-4H,5,13H2,1H3,(H,14,15)

InChI key

QEQDLKUMPUDNPG-UHFFFAOYSA-N

Application

AMCA is a useful agent for labeling proteins. Demonstrates a characteristic absorption peak at 345 nm with a fluorescence emission at 440-460 nm (blue region); however, when conjugated with proteins the absorption peak shifts to 355 nm with a fluorescence emission at 440-460 nm. It has a high fluorescence yield with a sufficient Stokes shift (100 nm) and is relatively photostable [1]. Relatively easy to activate and couple to the N-terminal amino group of a peptide resulting in an acid stable amide bond [2][3].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Useful blue fluorophore for immunofluorescence, λabs ~350 nm, λem ~440 nm[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Muraro et al.
The Journal of clinical investigation, 100(2), 339-349 (1997-07-15)
The pathogenesis of multiple sclerosis (MS) is currently ascribed in part to a T cell-mediated process targeting myelin components. The T cell response to one candidate autoantigen, myelin basic protein (MBP), in the context of HLA-DR15Dw2, has been previously studied
Meera Mohan et al.
British journal of haematology, 189(1), 67-71 (2019-12-11)
Gain of chromosome 1q21 and the gene expression-based GEP70 risk score are established prognostic markers for newly diagnosed Multiple Myeloma (MM) patients. Here we addressed the prognostic impact of these two markers in 81 relapsed/refractory (RR) MM patients treated with
D Delia et al.
Cytometry, 12(6), 537-544 (1991-01-01)
Antibodies coupled to 7-aminocoumarin (AMCA) emit a bright blue fluorescence under ultraviolet (UV) excitation and are therefore ideal for three-color immunofluorescence (IF) with fluorescein (FITC) and phycoerythrin (PE) labeled reagents; however, due to the different absorption spectra, the use of
S Callaci et al.
Molecular cell, 3(2), 229-238 (1999-03-17)
Luminescence resonance energy transfer measurements were used to show that binding of E. coli core RNA polymerase induced major changes in interdomain distances in the sigma 70 subunit. The simplest model describing core-induced changes in sigma 70 involves a movement
P M Nederlof et al.
Cytometry, 10(1), 20-27 (1989-01-01)
A method is described for visualizing three nucleic acid sequences simultaneously by in situ hybridization using a new blue immunofluorescent label, amino methyl coumarin acetic acid (AMCA), in combination with green and red fluorescing FITC and TRITC. Three chromosome-specific repetitive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service