Skip to Content
Merck

Skip To

07076

Castor Oil, Hydrogenated,Ethoxylated

Synonym(s):

Macrogolglycerol hydroxystearate, PEG-40 castor oil, Polyoxyl 40 hydrogenated castor oil, RH 40

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12161900
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

impurities

≤0.2% sulfated ash (verfied on random samples only), ≤1 ppm ethylene oxide, ≤10 ppm dioxan, ≤2 ppm arsenic (verfied on random samples only), ≤2.0% water, ≤5000 ppm acetic acid, ≤620 ppm diethylene glycol, ≤620 ppm ethylene glycol

ign. residue

≤0.25% (verfied onrandom samples only)

acid value

≤0.8 mg KOH/g

hydroxyl value

60‑75 mg KOH/g

iodine value

≤1.0 g I2/100 g

saponification value

50‑60 mg KOH/g

suitability

suitable for acidity or alkalinity

storage temp.

room temp

Application

Castor Oil, Hydrogenated,Ethoxylated is used to solubilize vitamins, water insoluble active substances and essential oils in water or mixtures of water and alcohol.

Analysis Note

congealing temperature 16-26°C

Other Notes

Castor Oil, Hydrogenated, Ethoxylated is the brand name for a range of non-ionic polyethoxylated detergents. It was developed for use as solubilizers and emulsifiers.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
30906C5135388920
description

-

description

-

description

pH range 6.0-8.0

description

-

suitability

suitable for acidity or alkalinity

suitability

-

suitability

-

suitability

-

storage temp.

room temp

storage temp.

-

storage temp.

-

storage temp.

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

impurities

≤2 ppm arsenic (verfied on random samples only), ≤0.2% sulfated ash (verfied on random samples only), ≤2.0% water, ≤620 ppm ethylene glycol, ≤5000 ppm acetic acid, ≤620 ppm diethylene glycol, ≤10 ppm dioxan, ≤1 ppm ethylene oxide

impurities

≤0.2% sulfated ash (verfied on random samples only), ≤ acetic acid (≤0.5 g/100g (%) verfied on random samples only), ≤0.1% oleic acid, ≤10 ppm dioxan (verfied on random samples only), alkalinity, complies, ≤1 ppm ethylene oxide (verfied on random samples only), ≤620 ppm ethylene glycol (verfied on random samples only), ≤0.1% palmitic acid, ≤620 ppm diethylene glycol (verfied on random samples only), ≤10 ppm heavy metals (verfied on random samples only), ≤0.5% water, ≤1.0% free fatty acids, ≤0.20% ricinoleic acid

impurities

-

impurities

-

acid value

≤0.8 mg KOH/g

acid value

≤2.0 mg KOH/g

acid value

-

acid value

-

ign. residue

≤0.25% (verfied onrandom samples only)

ign. residue

≤0.2%

ign. residue

-

ign. residue

-


Storage Class

11 - Combustible Solids

flash_point_f

467.6 °F

flash_point_c

242 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. What is the purity? Are there any doble-bonds left?

    1 answer
    1. The purity of this product is not determined. This product is a hydrogenated castor oil ethoxylate and residual double bonds are expected to be very low. The maximum allowable iodine value is 1.0, indicating minimal residual unsaturation. This information is lot-specific and reported in the product Certificate of Analysis. Please see the link below to access a sample or lot-specific Certificate:
      https://www.sigmaaldrich.com/product/sigma/07076#product-documentation

      Helpful?

  2. Good afternoon! What are the storage conditions for product 07076 (SIGMA), CAS 61788-85-0?

    1 answer
    1. The product may be stored at room temperature.

      Helpful?

Reviews

No rating value

Active Filters