Skip to Content
Merck
All Photos(4)

Key Documents

C7606

Sigma-Aldrich

Activated Charcoal

meets USP testing specifications

Synonym(s):

Activated charcoal, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

vapor pressure

<0.1 mmHg ( 20 °C)

form

powder, crystals or granules

autoignition temp.

842 °F

resistivity

1375 μΩ-cm, 20°C (graphite)

impurities

cyanogens, passes test
≤3.5% acid solubles

mp

3550 °C (lit.)

anion traces

chloride (Cl-): ≤0.2%
sulfate (SO42-): ≤0.2%
sulfide (S2-): passes test

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Activated charcoal has been used during the preparation of enzyme extract for β-1,3-glucanase and chitinase assay.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemical markers involved in horizontal resistance to Phytophthora infestans in potatoes.
Wolski EA, et al.
Fruit, Vegetable and Cereal Science and Biotechnology, 3, 89-94 (2009)
Kohsuke Ohmatsu et al.
Nature chemistry, 6(1), 47-51 (2013-12-19)
The development of a general catalytic method for the direct and stereoselective construction of contiguous all-carbon quaternary stereocentres remains a formidable challenge in chemical synthesis. Here, we report a highly enantio- and diastereoselective [3+2] annulation reaction of 5-vinyloxazolidinones and activated
Nathalie Butt et al.
Ecological applications : a publication of the Ecological Society of America, 23(4), 936-943 (2013-07-20)
A typical way to quantify aboveground carbon in forests is to measure tree diameters and use species-specific allometric equations to estimate biomass and carbon stocks. Using "citizen scientists" to collect data that are usually time-consuming and labor-intensive can play a
Yin Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(1), 533-538 (2013-12-25)
Stomatal pores surrounded by a pair of guard cells in the plant epidermis control gas exchange between plants and the atmosphere in response to light, CO2, and the plant hormone abscisic acid. Light-induced stomatal opening is mediated by at least
Scott Weichenthal et al.
Environmental research, 132, 305-310 (2014-05-20)
Increasing evidence suggests that ultrafine particles (UFPs) may contribute to cardiorespiratory morbidity. We examined the relationship between near road UFPs and several traffic and built environment factors to identify predictors that may be used to estimate exposures in population-based studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service