Skip to Content
Merck
All Photos(1)

Documents

71388

Sigma-Aldrich

Sodium chlorite

puriss. p.a., 80% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO2
CAS Number:
Molecular Weight:
90.44
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

Assay

80% (RT)

form

powder or crystals

pH

10-11 (20 °C, 100 g/L)

cation traces

Ag: ≤5 mg/kg
Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤1000 mg/kg
Li: ≤5 mg/kg
Mg: ≤10 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Tl: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].[O-]Cl=O

InChI

1S/ClHO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

UKLNMMHNWFDKNT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium chlorite is a sodium salt. Antimicrobial activity of acidified sodium chlorite on broiler carcasses has been reported in detail.

Application

Sodium chlorite may be used in the following studies:
  • Synthesis of chlorine dioxide.
  • Oxidation of primary alcohols to the corresponding aldehydes and of the aldehydes to the corresponding carboxylic acids.
  • As hydroxylating agent for the hydroxylation of androstenedione (steroid).

Other Notes

Reagent for the oxidation of ruthenium and molybdenum sulfide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1B - STOT RE 2

Target Organs

spleen

Supplementary Hazards

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Jia et al.
Advanced materials (Deerfield Beach, Fla.), 30(30), e1801347-e1801347 (2018-06-09)
Advanced textiles made of macroscopic fibers are usually prepared from synthetic fibers, which have changed lives over the past century. The shortage of petrochemical resources, however, greatly limits the development of the textile industry. Here, a facile top-down approach for
R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
Selective oxidation of aldehydes to carboxylic acids with sodium chlorite-hydrogen peroxide.
Dalcanale E and Montanari F.
The Journal of Organic Chemistry, 51(4), 567-569 (1986)
G K Kemp et al.
Journal of food protection, 63(8), 1087-1092 (2000-08-17)
An acidified sodium chlorite (ASC) solution was investigated for its antimicrobial effects on broiler carcasses processed under conditions similar to those used in U.S. commercial poultry facilities. Of particular interest was the ability of the ASC solution to reduce natural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service