Skip to Content
Merck
All Photos(1)

Key Documents

650439

Sigma-Aldrich

2,2,4-Trimethylpentane

HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%

Synonym(s):

Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC Plus
for residue analysis
Assay:
≥99.5%
technique(s):
HPLC: suitable
gas chromatography (GC): suitable
application(s):
food and beverages
bp:
98-99 °C (lit.)
vapor pressure:
41 mmHg ( 21 °C)
Pricing and availability is not currently available.

grade

HPLC Plus
for residue analysis

Quality Level

vapor density

3.9 (vs air)

vapor pressure

41 mmHg ( 21 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

745 °F

expl. lim.

6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

<0.01% water
<1.0 ppb Fluorescence (quinine) at 365 nm

evapn. residue

<0.0001%

halogenated residue

<10 ng/L (as heptachlor epoxide)

refractive index

n20/D 1.391 (lit.)

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

density

0.692 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 205 nm Amax: 1.00
λ: 225 nm Amax: 0.10
λ: 254 nm Amax: 0.01

application(s)

food and beverages

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for HPLC, spectrophotometry, environmental testing

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Liquid chromatography/electrospray mass spectrometric detection of fatty acid by charge reversal derivatization with more than 4-orders of magnitude improvement in sensitivity.
Bollinger JG, et al.
Journal of Lipid Research, 55, 603-603 (2013)
B G Short et al.
Toxicologic pathology, 14(2), 194-203 (1986-01-01)
Unleaded gasoline causes acute and chronic nephrotoxicity and renal tumors in male rats, but not female rats or mice of either sex. An active nephrotoxic component of unleaded gasoline has been identified as 2,2,4-trimethylpentane (TMP). The first objective of this
Physicochemical properties of liquid mixtures. 1. Viscosity, density, surface tension and refractive index of cyclohexane+ 2, 2, 4-trimethylpentane binary liquid systems from 25 C to 50 ?C.
Gomez-Diaz D, et al.
Journal of Chemical and Engineering Data, 46(3), 720-724 (2001)
Density and viscosity measurements of 2, 2, 4-trimethylpentane (isooctane) from 198 K to 348 K and up to 100 MPa.
Padua AAH, et al.
Journal of Chemical and Engineering Data, 41(6), 1488-1494 (1996)
E A Lock et al.
Toxicology and applied pharmacology, 91(2), 182-192 (1987-11-01)
Trimethylpentane (TMP) produces nephrotoxicity in male but not in female rats. The toxicity is characterized by an increase in protein droplets in proximal convoluted tubular cells and an increase in the renal concentration of the male-rat-specific protein alpha 2u-globulin. Subcellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service