Skip to Content
Merck
All Photos(2)

Documents

537535

Sigma-Aldrich

2-(2-Butoxyethoxy)ethyl acetate

≥99.2%

Synonym(s):

Butyldiglycol acetate, Diethylene glycol monobutyl ether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2OCH2CH2O(CH2)3CH3
CAS Number:
Molecular Weight:
204.26
Beilstein:
1771533
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21

Assay

≥99.2%

form

liquid

autoignition temp.

393 °F

expl. lim.

0.76 %, 135 °F
10.7 %, 203 °F

impurities

≤0.10% (water)

refractive index

n20/D 1.423 (lit.)

bp

245 °C (lit.)

mp

-32 °C

solubility

water: soluble 65 g/L at 20 °C

density

0.978 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCOCCOC(C)=O

InChI

1S/C10H20O4/c1-3-4-5-12-6-7-13-8-9-14-10(2)11/h3-9H2,1-2H3

InChI key

VXQBJTKSVGFQOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(2-Butoxyethoxy)ethyl acetate (Butyldiglycol acetate, BGDA) is a volatile organic compound that can be prepared by reacting diethylene glycol monobutyl ether with acetic acid using strongly acidic cationic exchange resin as catalyst.

Application

2-(2-Butoxyethoxy)ethyl acetate (Butyldiglycol acetate) may be used as a component to reduce the minimal film forming temperature (MTF) during poly(vinyl acetate)(PVAc) based composite film preparation. It may also be used during the formulation of thick film pastes of silver.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Composites of poly(vinyl acetate) filled with calcium carbonate: microscopy, diffractometry and thermophysical properties.
Kovacevic V, et al.
Polym. Eng. Sci., 39(8), 1433-1443 (1999)
Organic compounds in indoor air-their relevance for perceived indoor air quality?
Wolkoff P and Nielsen GD.
Atmospheric Environment, 35(26), 4407-4417 (2001)
Influence of surfactants treatment on silver powder and its thick films.
Rane SB, et al.
Materials Letters, 57(20), 3096-3100 (2003)
P J Deisinger et al.
Xenobiotica; the fate of foreign compounds in biological systems, 19(9), 981-989 (1989-09-01)
1. The in vitro hydrolysis of DGBA in rat blood and its in vivo metabolism and disposition in male Sprague-Dawley rats were studied. 2. DGBA (5 mM) was hydrolysed in rat blood to diethylene glycol monobutyl ether (DGBE) with a
Delayed and immediate hypersensitivity to carbitols.
T A Dawson et al.
Contact dermatitis, 21(1), 52-53 (1989-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service