Skip to Content
Merck
All Photos(3)

Key Documents

310093

Sigma-Aldrich

Magnesium hydroxide

reagent grade, 95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(OH)2
CAS Number:
Molecular Weight:
58.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent grade
form:
powder

grade

reagent grade

Quality Level

Assay

95%

form

powder

pH

9.5-10.5

mp

350 °C (lit.)

SMILES string

O[Mg]O

InChI

1S/Mg.2H2O/h;2*1H2/q+2;;/p-2

InChI key

VTHJTEIRLNZDEV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium hydroxide is a low corrosive inorganic compound that is widely used as an acid- and halogen-free flame retardant and smoke suppressant in various polymeric matrices in the plastics industry. It is a magnesium oxide intermediate that can be used in refractory, water and wastewater treatment, fuel gas desulfurization, pulp and paper industry, and as a fertilizer additive.

Application

Magnesium hydroxide can be used as:
  • A filler to prepare kenaf fiber-reinforced polymer composites.
  • A pH-adjusting agent in the production of succinic acid.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Flame retardant and mechanical properties of natural fibre-PP composites containing magnesium hydroxide.
Sain M, et al.
Polymer Degradation and Stability, 83(2), 363-367 (2004)
Synthesis of magnesium hydroxide and its calcinates by a precipitation method with the use of magnesium sulfate and poly (ethylene glycols).
Pilarska A, et al.
Powder Technology, 235, 148-157 (2013)
Magnesium hydroxide reinforced kenaf fibers/epoxy hybrid composites: Mechanical and thermomechanical properties
Saba N, et al.
Construction Building Materials, 201, 138-148 (2019)
Production of magnesium hydroxide from magnesium silicate for the purpose of CO2 mineralisation-Part 1: Application to Finnish serpentinite.
Nduagu E, et al.
Minerals Engineering, 30, 75-86 (2012)
Kinetics of magnesium hydroxide precipitation from sea bittern
Alamdari A, et al.
Chemical Engineering and Processing, 47, 215-221 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service