Skip to Content
Merck
All Photos(6)

Key Documents

248614

Sigma-Aldrich

Ammonium persulfate

Ammonium persulfate

ACS reagent, ≥98.0%

Synonym(s):

AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2S2O8
CAS Number:
Molecular Weight:
228.20
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
chunks or granules
powder or crystals
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

7.9 (vs air)

Assay

≥98.0%

form

chunks or granules
powder or crystals

reaction suitability

reagent type: oxidant

impurities

≤0.005% insolubles
≤0.04 meq/g Titr. acid

ign. residue

≤0.05%

pH

1-2 (25 °C, 228 g/L)

anion traces

Cl- and ClO4-: ≤0.001%

cation traces

Fe: ≤0.001%
Mn: ≤0.5 ppm
heavy metals (as Pb): ≤0.005%

SMILES string

N.N.OS(=O)(=O)OOS(O)(=O)=O

InChI

1S/2H3N.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h2*1H3;(H,1,2,3)(H,4,5,6)

InChI key

ROOXNKNUYICQNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium persulfateis a readily available oxidizing agent for various chemicaltransformations, and it is also usedas a bleaching agent in the chemical industry.

Application

Ammonium persulfate can be used as an oxidizing reagent for the solvent-free oxidation of:
  • Primary and secondary alcohols to corresponding carbonyl compounds in an aqueous medium.
  • Thiols to disulfides.
It can also be used as an initiator to initiate copolymerization of acrylonitrile and itaconic acid.
Catalyst for acrylamide gel polymerization.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

William H Thiel et al.
Methods in molecular biology (Clifton, N.J.), 1218, 187-199 (2014-10-17)
After a decade of work to address cellular uptake, the principal obstacle to RNAi-based therapeutics, there is now well-deserved, renewed optimism about RNAi-based drugs. Phase I and II studies have shown safe, strong, and durable-gene knockdown (80-90%, lasting for a
J M Brewer
Science (New York, N.Y.), 156(3772), 256-257 (1967-04-14)
Ammonium persulfate, a common polymerizing agent for acrylamide gels, can inactivate yeast enolase and produce increased electrophoretic heterogeneity during disc electrophoresis in gels containing 8M urea. The use of riboflavin and light for polymerization or thioglycolate for removal of the
Electrical conductivity response of polypyrrole to acetone vapor: effect of dopant anions and interaction mechanisms.
Ruangchuay L, et al.
Synthetic Metals, 140(1), 15-21 (2004)
S Pino et al.
Clinical chemistry, 42(2), 239-243 (1996-02-01)
The chloric acid method is most commonly used to obtain accurate and reproducible measurements of iodine and remove interfering substances. Unfortunately chloric acid is a potential hazard, requiring an explosion-proof hood, among other precautions. We have developed a simple, convenient
T Simonsson et al.
Nucleic acids research, 26(5), 1167-1172 (1998-04-04)
The c-myc oncogene is one of the most commonly malfunctioning genes in human cancers, and is an attractive target for anti-gene therapy. Although synthetic oligonucleotides designed to silence c-myc expression via one of its major control elements function well in

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service