Skip to Content
Merck
All Photos(4)

Key Documents

237051

Sigma-Aldrich

Aluminum chloride

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlCl3
CAS Number:
Molecular Weight:
133.34
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38120507
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
powder
Pricing and availability is not currently available.

grade

reagent

vapor pressure

1 mmHg ( 100 °C)

product line

ReagentPlus®

Assay

99%

form

powder

reaction suitability

reagent type: catalyst
core: aluminum

pH

2.4 (20 °C, 100 g/L)

mp

190 °C (lit.)

SMILES string

Cl[Al](Cl)Cl

InChI

1S/Al.3ClH/h;3*1H/q+3;;;/p-3

InChI key

VSCWAEJMTAWNJL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum chloride is an alcohol-soluble lewis acid.† Aluminum chloride mixed with sodium borohydride, in diglyme in a molar ratio affords a mixture having strong reducing properties.† It was reported to promote the Friedel-Crafts acylation reaction between 4-tert-butylbenzoyl chloride and mesitylene.†

Application

Aluminum chloride was used for the following studies:
  • Polymerization of benzene to afford p-polyphenyl.[1]
  • As a catalyst for the Friedel-Crafts acylation of buckymetallocenes with the acid chlorides.[2]
  • As a catalyst for the colorimetric microassay for the determination of lipid hydroperoxides.[3]

Features and Benefits

The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. Aluminum nitride can be synthesized from the elemnts with AlCl3 added to help facilitate crystallization.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of Alternative Oxidase Expression in Arabidopsis thaliana Protoplasts Treated with Aluminium.
Liu J, et al.
Bio-protocol, 5(13) (2015)
Yutaka Matsuo et al.
Dalton transactions (Cambridge, England : 2003), 43(20), 7407-7412 (2013-10-26)
Acylated buckyferrocene and ruthenocene, Fe(η(5)-C60Me5)(η(5)-C5H4COR) (R = Me, Ph, and CH[double bond, length as m-dash]CHPh) and Ru(η(5)-C60Me5)(η(5)-C5H4COR) (R = Me and Ph), were obtained by Friedel-Crafts acylation of the parent buckymetallocenes with the corresponding acid chlorides and aluminum chloride in
Shutao Wang et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(4), 1700712-1700712 (2018-05-04)
In the search for low-cost and large-scale stationary storage of electricity, nonaqueous aluminum chloride-graphite batteries (AlCl3-GBs) have received much attention due to the high natural abundances of their primary constituents, facile manufacturing, and high energy densities. Much research has focused
Rajkumar Kore et al.
Dalton transactions (Cambridge, England : 2003), 47(23), 7795-7803 (2018-06-01)
Two solids of differing Lewis acidities, triethylammonium tetrachlorozincate ([HN222]2[ZnCl4]) and AlCl3, have been combined across 0.1 to 0.9 mole fractions (with respect to [HN222]2[ZnCl4]), and homogeneous solid double salts or mixed metal [HN222]2x[(1 - x)AlCl3 + xZnCl4] double salt ionic
p-Polyphenyl from Benzene-Lewis Acid Catalyst-Oxidant. Reaction Scope and Investigation of the Benzene-Aluminum Chloride-Cupric Chloride System1.
Kovacic P and Oziomek J.
The Journal of Organic Chemistry, 29(1), 100-104 (1964)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service