Skip to Content
Merck
All Photos(5)

Documents

236500

Sigma-Aldrich

Sodium acetate trihydrate

ACS reagent, ≥99%

Synonym(s):

Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

≥99%

form

crystals or chunks

autoignition temp.

1112 °F

impurities

MnO4- reducers, passes test
≤0.005% insolubles

pH

7.5-9.2 (25 °C, 5%)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.005%
Fe: ≤5 ppm
K: ≤0.005%
Mg: ≤0.002%
heavy metals (as Pb): ≤5 ppm

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium acetate trihydrate (SAT) is a hydrated sodium salt of acetic acid. It is an excellent inorganic phase change material (PCM) due to its high energy storage density and thermal conductivity, widely used in the sector of dyeing, printing, pharmacy and buffering agent.

Application

Sodium acetate trihydrate can be used:
  • As a reducing agent in the synthesis of graphene nanosheets via chemical reduction of GO (graphene oxide) under moderate condition.
  • In the preparation of silicone/sodium acetate trihydrate hybrid.
  • As a catalyst in the solvent free acetylation of phenols, alcohols and thiols with acetic anhydride.

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methyl radical in irradiated single crystals of sodium acetate trihydrate.
Rogers MT and Kispert LD.
J. Chem. Phys. , 46(1), 221-223 (1967)
Water-responsive shape memory hybrid: Design concept and demonstration.
Fan K, et al.
Express Polymer Letters, 5(5), 409-416 (2011)
The crystal structure of sodium acetate trihydrate.
Cameron TS, et al.
Acta Crystallographica Section B, Structural Science, 32(1), 87-90 (1976)
Eagleson M.
Concise Encyclopedia Chemistry, 995-995 (1994)
Claire E Mason et al.
Journal of the American Society for Mass Spectrometry, 24(2), 274-285 (2013-01-04)
Single-stranded DNA-binding proteins (SSBs) are ubiquitous oligomeric proteins that bind with very high affinity to single-stranded DNA and have a variety of essential roles in DNA metabolism. Nanoelectrospray ionization mass spectrometry (nanoESI-MS) was used to monitor subunit exchange in full-length

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service