Skip to Content
Merck
All Photos(1)

Key Documents

234125

Sigma-Aldrich

Iron(III) ferrocyanide

Synonym(s):

Ferric ferrocyanide, Ferrihexacyanoferrate, Ferrocin, Prussian blue

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe4[Fe(CN)6]3
CAS Number:
Molecular Weight:
859.23
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

concentration

≥30% (ICP)

λmax

694 nm

SMILES string

[Fe+3].[Fe+3].[Fe+3].[Fe+3].N#C[Fe-4](C#N)(C#N)(C#N)(C#N)C#N.N#C[Fe-4](C#N)(C#N)(C#N)(C#N)C#N.N#C[Fe-4](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/18CN.7Fe/c18*1-2;;;;;;;/q;;;;;;;;;;;;;;;;;;3*-4;4*+3

InChI key

DNMNDNSFJMUUFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Iron(III) ferrocyanidecyclic can be used as:
  • A catalyst to synthesize cyclic carbonates by reaction between carbon dioxide and epoxides.
  • A precursor to prepare Fe-Fe3C nanoparticle encapsulated N-doped graphene layers via pyrolysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Y Vaheb et al.
Journal of nanoscience and nanotechnology, 12(11), 8710-8714 (2013-02-21)
We investigate the assembly of Prussian blue and Cs-Co-Cr Prussian blue analogue molecular nanomagnets into nano-patterned electrodes. Pd or Au gaps - 7-50 nm were fabricated on a SiO2/Si substrate using standard electron beam lithography and lift-off. Nanomagnets were positioned
Prussian blue and its analogues: electrochemistry and analytical applications.
Karyakin AA.
Electroanalysis, 13(10), 813-819 (2001)
Vasanthi Siruguri et al.
The Indian journal of medical research, 136(1), 89-97 (2012-08-14)
The present study was carried out on stored rice variety PAU 201 in Punjab that was not permitted for milling and public distribution due to the presence of damaged grains at levels exceeding the regulatory limits of 4.75 per cent.
Lijuan Han et al.
Biosensors & bioelectronics, 43, 120-124 (2013-01-08)
In this study, we report a triple-component sensor fabricated by freestanding graphite felt (GF), platinum nanoparticles (Pt) and Prussian blue (PB). Pt is ultrasonically-electrodeposited on GF to increase the conductivity and to render the catalysts to the chemical deposition of
Chen Zhai et al.
Biosensors & bioelectronics, 42, 124-130 (2012-12-04)
In this paper, chitosan-prussian blue-multiwall carbon nanotubes-hollow gold nanospheres (Chit-PB-MWNTs-HGNs) film was fabricated onto the gold electrode surface by one-step electrodeposition method; and then acetylcholinesterase (AChE) and Nafion were modified onto the film to prepare an AChE biosensor. Incorporating MWNTs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service