Skip to Content
Merck
All Photos(4)

Key Documents

215473

Sigma-Aldrich

Ammonium cerium(IV) nitrate

ACS reagent, ≥98.5%

Synonym(s):

CAN, Ceric ammonium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NH4)2(NO3)6
CAS Number:
Molecular Weight:
548.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥98.5%
grade:
ACS reagent
form:
powder or crystals

grade

ACS reagent

Quality Level

Assay

≥98.5%

form

powder or crystals

reaction suitability

reagent type: oxidant

impurities

≤0.05% insol. dil. H2SO4

anion traces

chloride (Cl-): ≤0.01%
phosphate (PO43-): ≤0.02%

cation traces

Fe: ≤0.005%

SMILES string

N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ce.2HNO3.4NO3.2H3N/c;6*2-1(3)4;;/h;2*(H,2,3,4);;;;;2*1H3/q+4;;;4*-1;;

InChI key

WIBGOERAEYJBOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Offers a novel route to tartronic acid derivatives via facile oxygenation of malonates.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective and efficient oxidation of sulfides to sulfoxides using ammonium cerium (IV) nitrate in the presence of a catalytic amount of KBr or NaBr.
Zolfigol MA, et al.
Monatshefte fur Chemie / Chemical Monthly, 139(8), 895--895- (2008)
Carbon-heteroatom bond-forming reactions mediated by cerium (IV) ammonium nitrate: An overview.
Nair V, et al.
Synlett, 14(2), 156-165 (2003)
Cerium (IV) ammonium nitrate (CAN) as a catalyst in tap water: A simple, proficient and green approach for the synthesis of quinoxalines.
More SV, et al.
Green Chemistry, 8(1), 91-95 (2006)
Polyethylene glycol (PEG) mediated green synthesis of 2, 5-disubstituted 1, 3, 4-oxadiazoles catalyzed by ceric ammonium nitrate (CAN).
Kidwai M, et al.
Green Chemistry Letters and Reviews, 31(1), 55-59 (2010)
A New. ALPHA.-Iodination of Ketones Using Iodine-Ammonium Cerium (IV) Nitrate in Alcohol or Acetic Acid.
Horiuchi CA and Kiji S.
Bulletin of the Chemical Society of Japan, 70(2), 421-426 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service