Skip to Content
Merck
All Photos(1)

Key Documents

199966

Sigma-Aldrich

Aluminum oxide

Brockmann I, standard grade, activated, acidic

Synonym(s):

Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352311
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
SB.52

Agency

suitable for EPA 1613

form

powder

manufacturer/tradename

CAMAG 504-C-I

technique(s)

ion chromatography: suitable

impurities

<0.03% Fe2O3
<0.03% SiO2
<0.4% Na2O

particle size

50-300 mesh

pore size

58 Å pore size

pH

4.5±0.5 ( in H2O)

mp

2040 °C (lit.)

anion traces

chloride (Cl-): ≤0.14 (mval/g)

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Brockmann aluminum oxides are suitable for column chromatography and adsorptive filtration.
The mechanism of formation of protective scales of α-Al2O3 (aluminum oxide) on surfaces of binary M-Al (M = Fe, Ni or Co) alloys at high temperatures has been studied. Activated aluminum oxides having different acidities and water concentrations (having Brockmann activity I-V) can easily detect various types of water (namely free water, pore water, and bound water) on the aluminum oxide present in soils.

Application

Aluminum oxide (Alumina) may be used for the removal of peroxides from the dehydration solvent (tetrahydrofuran, THF) in a protocol. Protocol involved the high resolution study of fine brain anatomy of fluorescently labeled mouse brains. It may be used in the fabrication of polycrystalline lithium iodide-Aluminum oxide solid electrolytes. The incorporation of aluminum oxide remarkably increased the conductivity of lithium iodide.
Aluminum oxide (acidic) can be used as an adsorbent for column chromatography. It may be used as an active surface for the acylation of aromatic ethers with carboxylic acids in the presence of trifluoroacetic anhydride under solvent-free conditions.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Efficient Syntheses of (a-Fluoropropargyl) phosphonate Esters.
Benayoud F, et al.
The Journal of Organic Chemistry, 61(15), 5159-5164 (1996)
A simple, efficient, and selective method for tetrahydropyranylation of alcohols on a solid phase of alumina impregnated with zinc chloride.
Ranu BC and Saha M.
The Journal of Organic Chemistry, 59(26), 8269-8270 (1994)
Micron-scale resolution optical tomography of entire mouse brains with confocal light sheet microscopy.
Silvestri L, et al.
Journal of Visualized Experiments, 80, e50696-e50696 (2013)
Conduction Characteristics of the Lithium Iodide-Aluminum Oxide Solid Electrolytes.
Liang CC.
Journal of the Electrochemical Society, 120(10), 1289-1292 (1973)
Sebastiaan P van Kessel et al.
Nature communications, 10(1), 310-310 (2019-01-20)
Human gut microbiota senses its environment and responds by releasing metabolites, some of which are key regulators of human health and disease. In this study, we characterize gut-associated bacteria in their ability to decarboxylate levodopa to dopamine via tyrosine decarboxylases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service