Skip to Content
Merck
All Photos(3)

Key Documents

13803

Sigma-Aldrich

Sulfur

99.5-100.5%, meets analytical specification of Ph. Eur., BP, USP, puriss., precipitated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
EC Number:
MDL number:
UNSPSC Code:
12141912
PubChem Substance ID:
NACRES:
SB.54
form:
solid
solubility:
carbon disulfide: in accordance 1 g/5mL
Pricing and availability is not currently available.

vapor density

8.9 (vs air)

vapor pressure

1 mmHg ( 183.8 °C)
10 mmHg ( 246 °C)

grade

puriss.

Assay

99.5-100.5%

form

solid

autoignition temp.

450 °F

quality

meets analytical specification of Ph. Eur., BP, USP
precipitated

resistivity

2E23 μΩ-cm, 20°C

impurities

acidity or alkalinity, complies
residual solvents, complies
≤0.01% free alkali (as NaOH)
≤0.02% free acid (as H2SO4)
≤0.5% water (Karl Fischer)

ign. residue

≤0.2% (as SO4)
≤0.3%

bp

444.7 °C (lit.)

mp

112.8 °C (rhombic) (lit.)
117-120 °C (lit.)
119.0 °C (monoclinic) (lit.)

solubility

carbon disulfide: in accordance 1 g/5mL

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg
sulfide (S2-): ≤10 mg/kg

cation traces

As: ≤40 mg/kg

suitability

complies for appearance of solution
corresponds for odor

SMILES string

[S]

InChI

1S/S

InChI key

NINIDFKCEFEMDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfur is present in abundant quantities in marine and freshwater sediments. It exists in the form of various sulfur compounds such as pyrite, acid volatile sulfides, elemental sulfur, sulfates and organic sulfur.Quantitative estimation of reduced inorganic sulfur compounds in modern sediments and shales by a chromium reduction technique has been described.
Sulfur is naturally present in cysteine and methionine amino acids, which can be detected by inductively coupled plasma mass spectrometry(ICP-MS).[1] It is also an important element required for plant growth, since it is useful in lowering the pH of the soil.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mccarty.B.L et al.
Fundamentals of Turfgrass and Agricultural Chemistry (2003)
Sulfur as the key element for quantitative protein analysis by capillary liquid chromatography coupled to element mass spectrometry
Wind M, et al.
Angewandte Chemie (International Edition in English), 42(29), 3425-3427 (2003)
Electron microscopy study of CVT grown Fe-sulphides.
Zavasnik J and Recnik A.
Journal of Crystal Growth, 367, 18-23 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service