Skip to Content
Merck
All Photos(1)

Documents

13767

Supelco

Silica gel orange

with moisture indicator free of heavy metals

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:

quality

with moisture indicator free of heavy metals

Quality Level

impurities

≤0.025% soluble in chloroform

loss

≤2% loss on drying, 150 °C, 3 h

particle size

2-5 mm

capacity

≥25 % absorption capacity (water)

bp

2230 °C

mp

>1600 °C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica gel orange (Drying pearls Orange) is a commonly employed drying agent.

Application

Silica gel orange (Drying pearls Orange) may be used to provide long-term storage conditions in the following studies:
  • Multiwell plates in a gel drying box during the solid-phase reverse transfection on cell microarrays.
  • Robust protocol for the reverse transfection of cells on small interfering (siRNA) arrays.
It was used to compose the spotting solution of transfection array.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Qingyu Zhang et al.
Brain, behavior, and immunity, 88, 471-481 (2020-04-14)
Increased kynurenine (Kyn) metabolized from tryptophan (Try) is a biomarker in the immune dysfunction of depression. However, the mechanism by which Kyn change promotes depression is poorly defined. Astrocytes are involved in the neuroinflammation of depression. Among the numerous inflammatory
Holger Erfle et al.
Methods in molecular biology (Clifton, N.J.), 785, 277-287 (2011-09-09)
Endocytosis is one of the most essential cellular processes, which enables cells to internalise diverse -material. It is crucial for regulation of receptor activity and signalling, cell polarisation, attachment and motility, and a great number of other cellular functions. A
Ren-Wei Du et al.
Experimental brain research, 238(11), 2549-2556 (2020-09-02)
Emerging evidence indicates that NLRP3 inflammasome-induced inflammation plays a crucial role in the pathogenesis of depression. Thus, inhibition of NLRP3 inflammasome activation may offer a therapeutic benefit in the treatment of depression. Metformin has been shown to have potential anti-inflammatory
Holger Erfle et al.
Journal of biomolecular screening, 13(7), 575-580 (2008-07-05)
Solid-phase reverse transfection on cell microarrays is a high-throughput method for the parallel transfection of mammalian cells. However, the cells transfected in this way have been restricted so far to microscopy-based analyses. Analysis methods such as reverse transcriptase-polymerase chain reaction
Miaomiao Jia et al.
The international journal of neuropsychopharmacology, 20(11), 886-895 (2017-10-11)
Leonurine is an active alkaloid that is extracted from Traditional Chinese Medicine Herba leonuri. Emerging evidence indicates that leonurine produces neuroprotective effects in ischemic stroke, Parkinson's disease, and Alzheimer's disease. However, the effect of leonurine in neuropsychiatric disorders, especially in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service