Skip to Content
Merck
All Photos(4)

Documents

13459

Sigma-Aldrich

Sodium metabisulfite

puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%

Synonym(s):

Sodium disulfite, Sodium pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O5
CAS Number:
Molecular Weight:
190.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

puriss.

Assay

97-100.5%

form

powder or crystals

quality

dry
meets analytical specification of Ph. Eur., BP, NF, FCC, E223

composition

SO2, 65.0-67.4%

impurities

residue of solvents, complies
≤0.02% thiosulfate (S2O3)

pH

3.5-5.0 (20 °C, 5%)

mp

>300 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
Hg: ≤1 mg/kg
Pb: ≤2 mg/kg
Se: ≤5 mg/kg
Zn: ≤10 mg/kg

suitability

complies for appearance of solution

SMILES string

[Na+].[Na+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2Na.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

HRZFUMHJMZEROT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium metabisulfite (MBS, Sodium disulfite) is an inorganic salt. It is the sodium salt of disulphurous (pyrosulfurous) acid. It is widely used as reducing agent in textile and photography industry. It is commonly added to various foods and wines as preservative.

Application

Sodium metabisulfite was used to initiate the cross-linking of polybutadiene in the cores of the vesicle membranes by free radical polymerization in solution. It has been used to prevent the oxidation (as antioxidant) during the preparation of stock solutions of 6-hydroxydopamine in various studies.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Johanne Louise Neiiendam et al.
Journal of neurochemistry, 91(4), 920-935 (2004-11-05)
The Neural Cell Adhesion Molecule (NCAM) plays a crucial role in development of the central nervous system regulating cell migration, differentiation and synaptogenesis. NCAM mediates cell-cell adhesion through homophilic NCAM binding, subsequently resulting in activation of the fibroblast growth factor
W Wright et al.
The American review of respiratory disease, 141(6), 1400-1404 (1990-06-01)
Sodium metabisulfite (MBS) is used as a preservative in food and wine and frequently triggers attacks of asthma. To determine the characteristics of responses to inhaled MBS, 30 asthmatic subjects and 16 nonasthmatic subjects inhaled MBS, in concentrations of 6.2
Zhiliang Cheng et al.
Langmuir : the ACS journal of surfaces and colloids, 24(15), 8169-8173 (2008-06-24)
The ability of chelated Gd to serve as an effective magnetic resonance (MR) contrast agent largely depends on fast exchange rates between the Gd-bound water molecules and the surrounding bulk water. Because water diffuses slowly across lipid bilayers, liposomes with
Laurie H L Sellings et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 23(15), 6295-6303 (2003-07-18)
Convergent evidence suggests that amphetamine (AMPH) exerts its rewarding and locomotor stimulating effects via release of dopamine in the nucleus accumbens. However, there is no consensus as to the relative contributions of core and medial shell subregions to these effects.
Eagleson M.
Concise Encyclopedia Chemistry, 337-337 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service