13438
Sodium bisulfite solution
purum, ~40%
Synonym(s):
Sodium hydrogen sulfite
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NaHSO3
CAS Number:
Molecular Weight:
104.06
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
liquid
Recommended Products
grade
purum
Quality Level
form
liquid
concentration
~40%
SMILES string
[Na+].OS([O-])=O
InChI
1S/Na.H2O3S/c;1-4(2)3/h;(H2,1,2,3)/q+1;/p-1
InChI key
DWAQJAXMDSEUJJ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Quantitative reaction of sodium bisulphite with uracil and cytosine at pH 6 has been investigated.
Sodium bisulfite solution can be used in isolating carbonyl compounds like aldehydes, via derivatization, followed by solvent extraction.
Sodium bisulfite solution can be used in isolating carbonyl compounds like aldehydes, via derivatization, followed by solvent extraction.
Application
Sodium bisulfite solution may be used for the production of SO3- radicals, by reaction with OH in irradiated aqueous solutions containing N2O. Chemistry of these radicals has been investigated by pulse radiolysis with kinetic spectrophotometry.
Other Notes
The article number 13438-4X2.5L-R will be discontinued. Please order the single bottle 13438-2.5L-R which is physically identical with the same exact specifications.
The article number 13438-6X1L-R will be discontinued. Please order the single bottle 13438-1L-R which is physically identical with the same exact specifications.
The article number 13438-6X1L-R will be discontinued. Please order the single bottle 13438-1L-R which is physically identical with the same exact specifications.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yufeng Ma et al.
International journal of environmental research and public health, 16(14) (2019-07-25)
Human exposure to polycyclic aromatic hydrocarbons (PAHs) results in adverse health implications. However, the specific impact of paternal preconception PAHs exposure has not been fully studied. In this study, a total of 219 men aged 24-53 were recruited and an
Hiroki Nakata et al.
Journal of anatomy, 235(2), 271-280 (2019-05-31)
The aim of the present study was to clarify the detailed morphology of efferent and epididymal ducts in adult mice using three-dimensional (3D) analysis. We reconstructed efferent and epididymal ducts in three adult mice using serial paraffin sections and high-performance
Armarego.F.L.W and Chai C
Purification of Laboratory Chemicals (2009)
Chemical behavior of sulfur trioxide (SO3-) and sulfur pentoxide (SO5-) radicals in aqueous solutions.
Huie RE and Neta P.
The Journal of Physical Chemistry, 88(23), 5665-5669 (1984)
Hironori Aoki et al.
Digestive diseases and sciences, 63(7), 1920-1928 (2018-03-17)
Colorectal serrated lesions (SLs) are important premalignant lesions whose clinical and biological features are not fully understood. We aimed to establish accurate colonoscopic diagnosis and treatment of SLs through evaluation of associations among the morphological, pathological, and molecular characteristics of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service