Skip to Content
Merck
All Photos(1)

Key Documents

Z2500000

Zolpidem tartrate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H42N6O2 · C4H6O6
CAS Number:
Molecular Weight:
764.87
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

zolpidem

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[n]21c(nc(c2CC(=O)N(C)C)c3ccc(cc3)C)ccc(c1)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O

InChI

1S/C19H21N3O.C4H6O6/c1-13-5-8-15(9-6-13)19-16(11-18(23)21(3)4)22-12-14(2)7-10-17(22)20-19;5-1(3(7)8)2(6)4(9)10/h5-10,12H,11H2,1-4H3;1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1

InChI key

NYVVVBWEVRSKIU-LREBCSMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Zolpidem tartrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marieke De Boeck et al.
Journal of pharmaceutical and biomedical analysis, 164, 57-61 (2018-10-22)
The analysis of biological samples, such as whole blood, comes with several sample preparation challenges. Biological matrices often contain a variety of endogenous components that can interfere with the determination of xenobiotics. Especially blood plasma proteins (e.g. serum albumin) are
Marieke De Boeck et al.
Forensic science international, 274, 44-54 (2017-01-18)
To date, thorough clean-up of complex biological samples remains an essential part of the analytical process. The solid phase extraction (SPE) technique is the well-known standard, however, its main weaknesses are the labor-intensive and time-consuming protocols. In this respect, dispersive
Xueyi Feng et al.
Drug testing and analysis, 11(7), 1076-1082 (2019-03-27)
Oral fluid zolpidem detection in the settings of drug-facilitated crime and roadside drug testing indicates recent zolpidem intake. Zolpidem pharmacokinetics in classical biological matrices such as blood and urine have been described; however, reports of such data based on oral
Xueyi Feng et al.
Journal of analytical toxicology, 41(9), 735-743 (2017-10-07)
A rapid, selective and sensitive LC-MS-MS method with a post-column addition of acetonitrile was developed and fully validated for the quantitative determination of zolpidem and its major metabolite, zolpidem phenyl-4-carboxylic acid (ZPCA), in oral fluid. Preliminary sample treatment was limited

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service