Z2376
Zinquin
≥95% (HPLC), solid
Synonym(s):
2-[[2-Methyl-8-[[(4-methylphenyl)sulfonyl]amino]-6-quinolinyl]oxy]acetic acid, Zinquin A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H18N2O5S
CAS Number:
Molecular Weight:
386.42
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
color
off-white
solubility
DMSO: soluble
methanol: soluble
functional group
carboxylic acid
storage temp.
2-8°C
SMILES string
Cc1ccc(cc1)S(=O)(=O)Nc2cc(OCC(O)=O)cc3ccc(C)nc23
InChI
1S/C19H18N2O5S/c1-12-3-7-16(8-4-12)27(24,25)21-17-10-15(26-11-18(22)23)9-14-6-5-13(2)20-19(14)17/h3-10,21H,11H2,1-2H3,(H,22,23)
InChI key
CGNKOBNGEFZRQU-UHFFFAOYSA-N
Application
Zinquin has been used to detect intracellular zinc.
Biochem/physiol Actions
Zinquin is a membrane-permeant fluorophore. It is used to detect zinc in cells by UV fluorescence. Zinquin ethyl ester can be used to monitor the change in intracellular concentrations of Zn2+ in thymocytes and hepatocytes. The ester is excited at 368nm and emits at 490nm.
Other Notes
Free acid or salt as available
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The supplementation of zinc increased the apoptosis of airway smooth muscle cells by increasing p38 phosphorylation.
Chiou Y L.
Environmental Toxicology and Pharmacology, 33(1), 70-77 (2012)
Chris D G and Joseph R L
Reviews in Fluorescence (2007)
Kilari Sreenivasulu et al.
Journal of food science, 75(4), H123-H128 (2010-06-16)
The effect of red wine (RW), red grape juice (RGJ), green tea (GT), and representative polyphenols on Caco-2 cell (65)Zn uptake was explored. RW, RGJ, and GT enhanced the uptake of zinc from rice matrix. Fractionation of RW revealed that
Kaya Lutter et al.
Molecular nutrition & food research, 54(2), 285-291 (2009-10-29)
3,3'-Dihydroxyisorenieratene (DHIR) is a structurally unusual carotenoid exhibiting bifunctional antioxidant properties. It is synthesized by Brevibacterium linens, used in dairy industry for the production of red smear cheeses. The compound protects cellular structures against photo-oxidative damage and inhibits the UV-dependent
Salmonella employs multiple mechanisms to subvert the TLR-inducible zinc-mediated antimicrobial response of human macrophages.
Kapetanovic R, et al.
Faseb Journal, 30(5), 1901-1912 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service