Skip to Content
Merck
All Photos(1)

Key Documents

Y0001658

Brimonidine tartrate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16BrN5O6
CAS Number:
Molecular Weight:
442.22
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

brimonidine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Brc1c2nccnc2ccc1NC3=NCCN3.O[C@H]([C@@H](O)C(=O)O)C(=O)O

InChI

1S/C11H10BrN5.C4H6O6/c12-9-7(17-11-15-5-6-16-11)1-2-8-10(9)14-4-3-13-8;5-1(3(7)8)2(6)4(9)10/h1-4H,5-6H2,(H2,15,16,17);1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1

InChI key

QZHBYNSSDLTCRG-LREBCSMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Brimonidine tartrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Schuster et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(33), 13538-13546 (2013-08-16)
We recently showed that spinal synergistic interactions between δ opioid receptors (δORs) and α2A adrenergic receptors (α2AARs) require protein kinase C (PKC). To identify which PKC isoforms contribute to analgesic synergy, we evaluated the effects of various PKC-isoform-specific peptide inhibitors
Sukru S Oner et al.
The Journal of biological chemistry, 288(33), 24091-24103 (2013-06-19)
Group II activators of G-protein signaling play diverse functional roles through their interaction with Gαi, Gαt, and Gαo via a G-protein regulatory (GPR) motif that serves as a docking site for Gα-GDP. We recently reported the regulation of the AGS3-Gαi
Mohammed M Ibrahim et al.
Pharmaceutical research, 30(11), 2818-2831 (2013-06-19)
Preparation of topical ophthalmic formulations containing brimonidine-loaded nanoparticles prepared from various biodegradable polymers-PCL, PLA and PLGA-for sustained release of brimonidine as a once daily regimen for management of glaucoma. Nanoparticles were prepared using spontaneous emulsification solvent diffusion method then characterized
Jie Shen et al.
Investigative ophthalmology & visual science, 55(2), 1056-1066 (2014-01-23)
We compared ocular and systemic pharmacokinetics of brimonidine and dexamethasone following a single intravitreal dose in animals with blood-retinal barrier (BRB) breakdown and in healthy controls. We induced BRB breakdown in rabbits by intravitreal injection of recombinant human VEGF165 and
T Realini et al.
Eye (London, England), 27(7), 841-847 (2013-05-04)
To describe pooled efficacy and safety data from two phase 3 studies comparing brinzolamide 1%/brimonidine 0.2% fixed combination (BBFC) with its component medications, brinzolamide and brimonidine, in patients with open-angle glaucoma or ocular hypertension. Data were pooled from two nearly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service