Skip to Content
Merck
All Photos(1)

Key Documents

Y0001604

Finasteride for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Finasteride, N-(2-methyl-2-propyl)-3-oxo-4-aza-5α-androst-1-ene-17β-carboxamide, N-tert-Butyl-3-oxo-4-aza-5α-androst-1-en-17β-carboxamide, MK-906

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H36N2O2
CAS Number:
Molecular Weight:
372.54
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

finasteride

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](C(=O)NC(C)(C)C)[C@@]4(C)CC[C@]3([H])[C@@]1(C)C=CC(=O)N2

InChI

1S/C23H36N2O2/c1-21(2,3)25-20(27)17-8-7-15-14-6-9-18-23(5,13-11-19(26)24-18)16(14)10-12-22(15,17)4/h11,13-18H,6-10,12H2,1-5H3,(H,24,26)(H,25,27)/t14-,15-,16-,17+,18+,22-,23+/m0/s1

InChI key

DBEPLOCGEIEOCV-WSBQPABSSA-N

Gene Information

human ... SRD5A2(6716)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Finasteride for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Selective 5α-reductase inhibitor; antiandrogen.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah A Finn et al.
CNS drug reviews, 12(1), 53-76 (2006-07-13)
Finasteride is the first 5alpha-reductase inhibitor that received clinical approval for the treatment of human benign prostatic hyperplasia (BPH) and androgenetic alopecia (male pattern hair loss). These clinical applications are based on the ability of finasteride to inhibit the Type
D Berthold et al.
Andrologia, 44 Suppl 1, 836-837 (2011-09-29)
Leydig cell tumours (LCTs) of the testis are rare. Their origin is still unknown. This case report describes a potential relationship between LCT and prolonged exposure to Finasteride.
Souhil Lebdai et al.
BJU international, 105(4), 456-459 (2009-11-26)
Prostate cancer is the most common neoplasm in men. Encouraging results are emerging in prostate cancer risk reduction with 5alpha-reductase (5AR) inhibitors. The Prostate Cancer Prevention Trial (PCPT) showed that prostate cancer risk is reduced by finasteride. However, there was
Aditya K Gupta et al.
The Journal of dermatological treatment, 25(2), 156-161 (2013-06-19)
In the light of post-marketing reports of persistent sexual dysfunction with the use of finasteride, analysis of the extent of risk associated with 5α-reductase inhibitor treatment for androgenetic alopecia (AGA) is warranted. This study sought to evaluate the efficacy of
José Manuel Mella et al.
Archives of dermatology, 146(10), 1141-1150 (2010-10-20)
Androgenetic alopecia is the most common form of alopecia in men. To determine the efficacy and safety of finasteride therapy for patients with androgenetic alopecia. MEDLINE, EMBASE, CINAHL, Cochrane Registers, and LILACS were searched for randomized controlled trials reported in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service