Skip to Content
Merck
All Photos(1)

Key Documents

Y0001358

Glycopyrronium impurity N

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Glycopyrrolate, α-cyclopentylmandelic acid ester with 3-hydroxy-1,1-dimethylpyrrolidinium bromide, 1-methyl-3-pyrrolidyl α-cyclopentylmandelate methobromide, 1-methyl-3-pyrrolidyl α-phenyl-α-cyclopentylglycolate methobromide, 3-(2-phenyl-2-cyclopentylglycoloyloxy)-1,1-dimethylpyrrolidinium bromide, 3-hydroxy-1,1-dimethylpyrrolidinium bromide α-cyclopentylmandelate, AHR-504, Copyrrolate, Gastrodyn, NSC 250836, NSC 251251, NSC 251252, NVA-237, glycopyrronium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28NO3 · Br
CAS Number:
Molecular Weight:
398.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

glycopyrronium

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Br-].C[N+]1(C)CCC(C1)OC(=O)C(O)(C2CCCC2)c3ccccc3

InChI

1S/C19H28NO3.BrH/c1-20(2)13-12-17(14-20)23-18(21)19(22,16-10-6-7-11-16)15-8-4-3-5-9-15;/h3-5,8-9,16-17,22H,6-7,10-14H2,1-2H3;1H/q+1;/p-1

InChI key

VPNYRYCIDCJBOM-UHFFFAOYSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Glycopyrronium impurity N EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Glycopyrrolate is long-acting muscarinic antagonist (LAMA). It is kinetically selective muscarinic M3 receptor antagonist.
Long-acting muscarinic antagonist (LAMA); anticholinergic; antispasmodic; preanesthetic medicant

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

F Paton et al.
British journal of anaesthesia, 105(5), 558-567 (2010-10-12)
The cost-effectiveness of sugammadex for the routine reversal of muscle relaxation produced by rocuronium or vecuronium in UK practice is uncertain. We performed a systematic review of randomized controlled trials of sugammadex compared with neostigmine/glycopyrrolate and an economic assessment of
M A Valley et al.
Regional anesthesia, 19(6), 423-428 (1994-11-01)
Penile erections following the initiation of either axial regional or general anesthesia is rare; however, when it occurs in patients undergoing urologic procedures it may delay, or even cancel, the planned surgery. The purpose of this case report and review
Nada Ghoneim et al.
PloS one, 9(9), e106888-e106888 (2014-09-23)
Enteral formula feeding is a risk factor for necrotizing enterocolitis (NEC) in premature infants, yet studies are conflicting regarding the safest timing for introduction and advancement of feeds. Our aim was to test the effects of early vs. late initiation
E N Schachter
Drugs of today (Barcelona, Spain : 1998), 49(7), 437-446 (2013-08-06)
Chronic obstructive pulmonary disease (COPD) is a worldwide problem causing prolonged and progressive morbidity as well as premature mortality. Pharmacologic treatment consists primarily in the relief of symptoms and preventing or minimizing the consequences of exacerbations. Central to the pharmacologic
Glycopyrrolate.
R K Mirakhur et al.
General pharmacology, 12(6), 423-427 (1981-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service