Skip to Content
Merck
All Photos(1)

Key Documents

Y0001327

Atorvastatin calcium trihydrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Atorvastatin hemicalcium salt sesquihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H68CaF2N4O10 · 3H2O
CAS Number:
Molecular Weight:
1209.39
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

atorvastatin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/2C33H35FN2O5.Ca.3H2O/c2*1-21(2)31-30(33(41)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)36(31)18-17-26(37)19-27(38)20-28(39)40;;;;/h2*3-16,21,26-27,37-38H,17-20H2,1-2H3,(H,35,41)(H,39,40);;3*1H2/q;;+2;;;/p-2/t2*26-,27-;;;;/m11..../s1

InChI key

SHZPNDRIDUBNMH-NIJVSVLQSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Atorvastatin calcium trihydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hongshi Li et al.
Cardiovascular journal of Africa, 30(5), 268-274 (2019-07-31)
Our previous experiments showed that the transient sodium current (INa) was abnormally increased in early ischaemia and atorvastatin could inhibit INa. The aim of this study was to observe the time-dependent effects of simulated ischaemia on INa and characterise the
Fang Hu et al.
AMB Express, 9(1), 37-37 (2019-03-20)
The aim of the present study was to evaluate the protective effects of combined atorvastatin and amygdalin in a rat model of endometriosis. Tumor necrosis factor-α (TNF-α), interleukin-6 (IL-6), matrix metalloproteinase-2 (MMP-2) and MMP-9 levels in the peritoneal fluid were
Hui-Hui Li et al.
Acta pharmacologica Sinica, 41(3), 327-335 (2019-07-31)
23,24-Dihydrocucurbitacin B (designated as C95 in this article) is a cucurbitane triterpenoid that has been shown to possess a variety of pharmacological activities, such as anti-inflammatory and anti-HIV-1 activities etc. In this study, we investigated the effects of 23,24-dihydrocucurbitacin B
Masaki Watanabe et al.
Drug metabolism and pharmacokinetics, 30(2), 198-204 (2015-05-21)
Since drug-drug interaction (DDI) can affect organic anion-transporting polypeptide (OATP) and cause clinical events, prediction of such DDI is important in early clinical development. Although statins are useful probes for OATP-mediated DDI, endogenous probes would be more practical for predicting
Alexander Treiber et al.
The Journal of pharmacology and experimental therapeutics, 350(1), 130-143 (2014-04-29)
Treatment of pulmonary arterial hypertension with the endothelin receptor antagonist bosentan has been associated with transient increases in liver transaminases. Mechanistically, bosentan inhibits the bile salt export pump (BSEP) leading to an intrahepatic accumulation of cytotoxic bile salts, which eventually

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service