Skip to Content
Merck
All Photos(1)

Key Documents

Y0001210

Tobramycin for identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Tobramycin, Nebramycin Factor 6, O-[3-Amino-3-deoxy-α-D-glucopyranosyl-(1→6)]-O-[2,6-diamino-2,3,6-trideoxy-α-D-ribohexopyranosyl-(1→4)]-2-deoxy-D-streptamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H37N5O9
CAS Number:
Molecular Weight:
467.51
Beilstein:
1357507
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tobramycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@@H](N)[C@H]3O)[C@H]2O)[C@H](N)C[C@@H]1O

InChI

1S/C18H37N5O9/c19-3-9-8(25)2-7(22)17(29-9)31-15-5(20)1-6(21)16(14(15)28)32-18-13(27)11(23)12(26)10(4-24)30-18/h5-18,24-28H,1-4,19-23H2/t5-,6+,7+,8-,9+,10+,11+,12+,13+,14-,15+,16-,17+,18+/m0/s1

InChI key

NLVFBUXFDBBNBW-SNGYORCQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tobramycin for identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Tobramycin is an aminoglycoside.
Mode of Action: Binds to 70S ribosomal subunit; inhibits translocation; elicits miscoding.
Spectrum of Activity: Gram negative bacteria. Not effective against Enterococci.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Foucaud et al.
Revue de pneumologie clinique, 58(3 Pt 1), 131-138 (2002-12-19)
Aerosol delivery of antibiotics offers the potential to achieve high antibiotic concentrations at the site of infection while reducing the risk of systemic untoward effects because of minimal resorption into the bloodstream. We reviewed knowledge acquired in this field over
A G Chermenskiĭ et al.
Terapevticheskii arkhiv, 82(8), 76-78 (2010-09-29)
The results of a few studies evaluating the efficacy and safety of nebulized tobramycin solution used in patients with cystic fibrosis have been published to date. Pulmonary deposition of inhaled tobramycin and its lung produced concentrations are rather high (10-25
P J Gavin et al.
Diagnostic microbiology and infectious disease, 47(2), 427-430 (2003-10-03)
Medical treatment alone is rarely successful in left-sided infective endocarditis caused by Pseudomonas aeruginosa. We report the cure of such a case with high-dose meropenem in combination with tobramycin.
Jennifer K Hagerman et al.
Expert opinion on pharmacotherapy, 8(4), 467-475 (2007-02-21)
Cystic fibrosis patients suffer increased sputum production and a notable decline in respiratory function throughout the progression of their disease. Patients are left vulnerable to respiratory colonization/infection from a number of pathogens, including Pseudomonas aeruginosa. At present, the only antibiotic
Michael D Parkins et al.
Expert review of respiratory medicine, 5(5), 609-622 (2011-10-01)
Lung disease in cystic fibrosis (CF) is typified by the development of chronic airways infection culminating in bronchiectasis and progression to end-stage respiratory disease. Pseudomonas aeruginosa, a ubiquitous gram-negative bacteria, is the archetypical CF pathogen and is associated with an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service