Skip to Content
Merck
All Photos(1)

Key Documents

Y0001133

Docetaxel trihydrate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H53NO14 · 3H2O
CAS Number:
Molecular Weight:
861.93
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

docetaxel

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N([C@H]([C@@H](O)C(=O)O[C@H]2C[C@]3([C@H]([C@@H]5[C@]6([C@H](OC6)C[C@@H]([C@]5(C(=O)[C@@H](C(=C2C)C3(C)C)O)C)O)OC(=O)C)OC(=O)c4ccccc4)O)c1ccccc1)C(=O)OC(C)(C)C.O.O.O

InChI

1S/C43H53NO14.3H2O/c1-22-26(55-37(51)32(48)30(24-15-11-9-12-16-24)44-38(52)58-39(3,4)5)20-43(53)35(56-36(50)25-17-13-10-14-18-25)33-41(8,34(49)31(47)29(22)40(43,6)7)27(46)19-28-42(33,21-54-28)57-23(2)45;;;/h9-18,26-28,30-33,35,46-48,53H,19-21H2,1-8H3,(H,44,52);3*1H2/t26-,27-,28+,30-,31+,32+,33-,35-,41+,42-,43+;;;/m0.../s1

InChI key

XCDIRYDKECHIPE-QHEQPUDQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Docetaxel trihydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuji Mukai et al.
Biological & pharmaceutical bulletin, 40(7), 1078-1085 (2017-07-05)
Fluvastatin, which is one of the hydroxymethylglutaryl-CoA (HMG-CoA) reductase inhibitors (statins), is primarily metabolized by CYP2C9 and to a lesser extent by CYP3A4 and CYP2C8. Predictions of drug-drug interactions (DDI) are important for the safety of combination therapies with statins
Valentina Guarneri et al.
Breast cancer research and treatment, 122(1), 181-188 (2010-04-03)
Long-term bisphosphonate therapy is associated with increased risk of osteonecrosis of the jaw (ONJ). In a retrospective analysis, a 16% ONJ incidence was reported in patients receiving bisphosphonates with anti-angiogenic therapy (bevacizumab or sunitinib) for bone metastases from breast, colon
Ryan M Franke et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 28(30), 4562-4567 (2010-09-22)
To assess whether the low incidence of severe neutropenia in castrated men with prostate cancer treated with docetaxel is the result of changes in systemic clearance. A total of 10 noncastrated and 20 castrated men with prostate cancer were studied
R Greil et al.
European journal of surgical oncology : the journal of the European Society of Surgical Oncology and the British Association of Surgical Oncology, 35(10), 1048-1054 (2009-03-03)
To evaluate the triplet combination of bevacizumab, capecitabine and docetaxel (XTA) as neoadjuvant therapy for breast cancer. Patients with invasive, HER2-negative, nonmetastatic breast cancer (T2-4c >2cm) and no prior systemic therapy received six 21-day cycles of XTA (bevacizumab 15mg/kg, day
David W Miles et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 28(20), 3239-3247 (2010-05-26)
The efficacy and safety of combining bevacizumab (7.5 and 15 mg/kg) with docetaxel as first-line therapy for human epidermal growth factor receptor 2 (HER2) -negative, locally recurrent or metastatic breast cancer (MBC) was investigated in a three-arm, placebo-controlled, phase III

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service