Skip to Content
Merck
All Photos(1)

Key Documents

Y0001080

Meloxicam

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13N3O4S2
CAS Number:
Molecular Weight:
351.40
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

meloxicam

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C14H13N3O4S2/c1-8-7-15-14(22-8)16-13(19)11-12(18)9-5-3-4-6-10(9)23(20,21)17(11)2/h3-7,19H,1-2H3,(H,15,16)/b13-11+

InChI key

DWMREKMVXIFPFM-ACCUITESSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Meloxicam EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cara L Field et al.
Journal of avian medicine and surgery, 26(4), 232-238 (2013-02-16)
A 22-year-old female African black-footed penguin (Spheniscus demersus), housed indoors with other African and rockhopper penguins, was presented acutely with lethargy, ataxia, and hind limb weakness after a molt. The penguin would assume a hunched position and, when resting, sat
Piotr Dorniak et al.
Endocrinology, 154(2), 931-941 (2012-12-25)
During early pregnancy in sheep, the elongating conceptus secretes interferon-τ (IFNT) and the conceptus as well as endometrial epithelia produce prostaglandins (PG) via PG synthase 2 (PTGS2) and cortisol via hydroxysteroid (11-β) dehydrogenase 1 (HSD11B1). Ovarian progesterone induces and PG
Heidi L Hoefer
Compendium (Yardley, PA), 35(2), E5-E5 (2013-03-28)
A 2-year-old, intact female guinea pig presented to the clinic for a history of squealing and straining to urinate for 2 days. The husbandry of this animal was acceptable, and the diet consisted of commercial alfalfa-based guinea pig pellets, alfalfa
Tomasz Maślanka et al.
Veterinary immunology and immunopathology, 151(3-4), 248-262 (2012-12-26)
In view of the lack of data on the effect of meloxicam (non-steroidal anti-inflammatory drug) on bovine γδ T cells (WC1(+) cells) and very poorly recognized effects of dexamethasone (steroidal anti-inflammatory drug) on these cells, the purpose of the present
Milko Radicioni et al.
International journal of clinical pharmacology and therapeutics, 51(3), 234-243 (2013-02-06)
Meloxicam is a non-steroidal anti-inflammatory drug, indicated for osteoarthritis exacerbations, rheumatoid arthritis and ankylosing spondylitis symptomatic treatment. To compare the bioavailability of a 15 mg meloxicam orodispersible tablet (ODT) and a reference 15 mg tablet in healthy volunteers. Two randomized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service