Skip to Content
Merck
All Photos(1)

Documents

Y0001047

Teicoplanin for identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Teicoplanin, Targocid, Teichomycin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

teicoplanin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

InChI

1S/C88H97Cl2N9O33/c1-3-4-5-6-7-8-9-10-60(108)94-68-74(113)71(110)58(32-101)129-87(68)132-78-55-26-40-27-56(78)126-52-18-14-38(24-47(52)90)77(131-86-67(92-34(2)103)73(112)70(109)57(31-100)128-86)69-84(121)98-66(85(122)123)45-29-42(105)30-54(127-88-76(115)75(114)72(111)59(33-102)130-88)61(45)44-23-37(13-15-49(44)106)63(81(118)99-69)96-83(120)65(40)97-82(119)64-39-21-41(104)28-43(22-39)124-53-25-36(12-16-50(53)107)62(91)80(117)93-48(79(116)95-64)20-35-11-17-51(125-55)46(89)19-35/h11-19,21-30,48,57-59,62-77,86-88,100-102,104-107,109-115H,3-10,20,31-33,91H2,1-2H3,(H,92,103)(H,93,117)(H,94,108)(H,95,116)(H,96,120)(H,97,119)(H,98,121)(H,99,118)(H,122,123)/t48-,57-,58-,59-,62-,63-,64+,65-,66-,67-,68-,69+,70-,71-,72-,73-,74-,75+,76+,77-,86+,87+,88+/m1/s1

InChI key

BJNLLBUOHPVGFT-QRZIFLFXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Teicoplanin for identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Teicoplanin is a glycopeptide antibiotic produced by Actinoplanes teichomiceticus that is effective against Gram-positive bacteria resistant to β-lactam antibiotics. Its mechanism of action is through inhibition of bacterial cell wall biosynthesis.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuli Svetitsky et al.
Antimicrobial agents and chemotherapy, 53(10), 4069-4079 (2009-07-15)
Vancomycin and teicoplanin are the glycopeptides currently in use for the treatment of infections caused by invasive beta-lactam-resistant gram-positive organisms. We conducted a systematic review and meta-analysis of randomized controlled trials that have compared vancomycin and teicoplanin administered systemically for
Michael D Nailor et al.
Infectious disease clinics of North America, 23(4), 965-982 (2009-11-17)
An overview of the mechanism of action, dosing, clinical indications, and toxicities of the glycopeptide vancomycin is provided. The emerging gram-positive bacterial resistance to antimicrobials and its mechanisms are reviewed. Strategies to control this emergence of resistance are expected to
[Glycopeptides (vancomycin, teicoplanin)].
Itaru Furuta et al.
Nihon rinsho. Japanese journal of clinical medicine, 62 Suppl 12, 423-426 (2005-01-22)
Edith Carbone et al.
Medicina, 62 Suppl 2, 25-29 (2002-12-17)
Teicoplanin is a glycopeptide antibiotic with similar spectrum to vancomycin. However, unlike this drug, teicoplanin can be administered by i.v. or i.m. route once daily thanks to its long half-life (88 to 182 hours). This pharmacokinetic characteristic is particularly interesting
H Jiang et al.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology, 32(9), 1121-1128 (2013-04-10)
Methicillin-resistant Staphylococcus aureus (MRSA) is an important cause of nosocomial pneumonia. Compared with glycopeptide antibiotics, linezolid achieves higher lung epithelial lining fluid concentrations, which may have an advantage in treating nosocomial pneumonia patients. The objective of this study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service