Skip to Content
Merck
All Photos(1)

Key Documents

Y0001020

Iopromide

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24I3N3O8
CAS Number:
Molecular Weight:
791.11
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

iopromide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(N(C)CC(O)CO)C1=C(I)C(NC(COC)=O)=C(I)C(C(NCC(CO)O)=O)=C1I

InChI

1S/C18H24I3N3O8/c1-24(4-9(28)6-26)18(31)12-13(19)11(17(30)22-3-8(27)5-25)14(20)16(15(12)21)23-10(29)7-32-2/h8-9,25-28H,3-7H2,1-2H3,(H,22,30)(H,23,29)

InChI key

DGAIEPBNLOQYER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iopromide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arthur Nasis et al.
Radiology, 272(3), 674-682 (2014-04-18)
To assess the long-term outcome and hospital readmission rate associated with a computed tomographic (CT) angiography-guided strategy used to examine patients who present to the emergency department (ED) with symptoms of possible acute coronary syndrome (ACS). The study was approved
Michel Habis et al.
Radiology, 274(3), 684-692 (2015-01-20)
To assess the feasibility of exercise perfusion computed tomography (CT) in patients suspected of having hemodynamically significant coronary stenosis. This study had institutional review board approval, and all patients gave informed consent. Thirty-two consecutive patients (26 men [mean age, 63
Ralph Gnannt et al.
Investigative radiology, 50(1), 17-23 (2014-09-10)
Mortality from abdominal abscesses ranges from 30% in treated cases up to 80% to 100% in patients with undrained or nonoperated abscesses. Various computed tomographic (CT) imaging features have been suggested to indicate infection of postoperative abdominal fluid collections; however
J Kooiman et al.
Journal of thrombosis and haemostasis : JTH, 12(10), 1658-1666 (2014-08-22)
Hydration to prevent contrast-induced acute kidney injury (CI-AKI) induces a diagnostic delay when performing computed tomography-pulmonary angiography (CTPA) in patients suspected of having acute pulmonary embolism. To analyze whether withholding hydration is non-inferior to sodium bicarbonate hydration before CTPA in
Chih-Jen Chou et al.
PloS one, 9(9), e106864-e106864 (2014-09-05)
Pigs are an optimal animal for conducting biomedical research because of their anatomical and physiological resemblance to humans. In contrast to the abundant resources available in the study of mice, few fluorescent protein-harboring porcine models are available for preclinical studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service