Skip to Content
Merck
All Photos(1)

Key Documents

Y0000851

Doxepin for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Doxepin hydrochloride, 11-(3-Dimethylaminopropylidene)-6,11-dihydrodibenz[b,e]oxepine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO · HCl
CAS Number:
Molecular Weight:
315.84
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

doxepin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CN(C)CC\C=C1/c2ccccc2COc3ccccc13

InChI

1S/C19H21NO.ClH/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;/h3-6,8-12H,7,13-14H2,1-2H3;1H/b17-11+;

InChI key

MHNSPTUQQIYJOT-SJDTYFKWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Doxepin for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan Lankford et al.
Sleep medicine, 13(2), 133-138 (2011-12-27)
The efficacy and safety of doxepin (DXP), a histamine H(1) receptor antagonist, was evaluated in elderly adults with sleep maintenance insomnia. This was a randomized, double-blind, placebo-controlled outpatient trial. Elderly adults meeting DSM-IV-TR criteria for primary insomnia were randomized to
Anna Maślanka et al.
Journal of AOAC International, 94(6), 1791-1799 (2012-02-11)
Stability of clonazepam, diazepam, haloperidol, and doxepin was determined in acidic solutions. In addition, determination of the kinetic and thermodynamic properties of this stability was carried out. Reaction rate constants (k), half-life times (t(0.1) and t(0.5)), and activation energy (Ea)
Andrew D Krystal et al.
Sleep, 34(10), 1433-1442 (2011-10-04)
To evaluate the efficacy and safety of doxepin (DXP) 3 mg and 6 mg in adults diagnosed with primary insomnia. The study was a randomized, double-blind, parallel-group, placebo-controlled trial. Patients meeting DSM-IV-TR criteria for primary insomnia were randomized to 35
Hancheng Cai et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 14(5), 546-552 (2012-01-10)
(11)C-Doxepin is an established positron emission tomography (PET) probe for imaging the histamine H1 receptor, which is associated with various neurological disorders and allergic diseases. A fully automated current Good Manufacturing Practices (cGMP)-compliant radiosynthesis is therefore desirable in order to
Christopher J Jones et al.
Analytical chemistry, 84(16), 7099-7106 (2012-08-03)
The capillary isotachophoresis (cITP) separation of the isomers of the tricyclic antidepressant doxepin using β-cyclodextrin (β-CD) as a buffer additive is investigated by online microcoil NMR detection. Capillary electrophoresis (CE) is also used to determine the binding constant between the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service