Skip to Content
Merck
All Photos(1)

Key Documents

Y0000750

Dorzolamide for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dorzolamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O4S3 · HCl
CAS Number:
Molecular Weight:
360.90
Beilstein:
5896026
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dorzolamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CCN[C@H]1C[C@H](C)S(=O)(=O)c2sc(cc12)S(N)(=O)=O

InChI

1S/C10H16N2O4S3.ClH/c1-3-12-8-4-6(2)18(13,14)10-7(8)5-9(17-10)19(11,15)16;/h5-6,8,12H,3-4H2,1-2H3,(H2,11,15,16);1H/t6-,8-;/m0./s1

InChI key

OSRUSFPMRGDLAG-QMGYSKNISA-N

Gene Information

human ... CA2(760)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dorzolamide for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2

Target Organs

Central nervous system,Gastrointestinal tract,Bone,Blood,Bladder

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuhiro Ikeda et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 250(6), 809-814 (2012-01-05)
Cystoid macular edema (CME) is one of the common complications of retinitis pigmentosa (RP), and is responsible for patient complications such as blurred and reduced visual acuity and for subsequent atrophic changes in the fovea. The objective of this work
Ikuyo Ohguro et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 28(4), 392-396 (2012-04-07)
The purpose of the present study is to evaluate the effects of a fixed combination of 0.5% timolol maleate (TM) and 1% dorzolamide hydrochloride (DZ) (FCTD(1%)) on optic nerve head (ONH) blood circulation. A drop of 0.5% TM, 1% DZ
Vikas Gulati et al.
Archives of ophthalmology (Chicago, Ill. : 1960), 130(6), 677-684 (2012-02-15)
To evaluate the interaction of intraocular pressure(IOP)–lowering medications with physiologic day and night changes in aqueous humor dynamics in participants with ocular hypertension. Thirty participants were enrolled in thisdouble-masked, randomized, crossover study. Each participant underwent aqueous humor dynamics measurements at
Yasuhiro Ikeda et al.
The British journal of ophthalmology, 97(9), 1187-1191 (2013-06-21)
To evaluate the therapeutic effect of continuous treatment with topical dorzolamide (a carbonic anhydrase inhibitor) for cystoid macular oedema (CME) associated with retinitis pigmentosa (RP). 18 eyes in 10 patients with CME secondary to RP were included. Baseline visual acuity
Mohamed A Genead et al.
Retina (Philadelphia, Pa.), 32(4), 826-833 (2011-09-03)
: To determine the value of a topical carbonic anhydrase inhibitor on the macular thickness and function in choroideremia patients with cystoid macular edema. : Two choroideremia patients with cystoid macular edema, observed by spectral-domain optical coherence tomography, were treated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service