Skip to Content
Merck
All Photos(1)

Key Documents

Y0000721

Mirtazapine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Mirtazapine, 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3
CAS Number:
Molecular Weight:
265.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mirtazapine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1CCN2C(C1)c3ccccc3Cc4cccnc24

InChI

1S/C17H19N3/c1-19-9-10-20-16(12-19)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)20/h2-8,16H,9-12H2,1H3

InChI key

RONZAEMNMFQXRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mirtazapine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Mirtazapine is a noradrenergic and specific serotonergic antidepressant (NaSSA). Mirtazapine agonizes selective adrenergic and serotonergic receptors so that both NE release and 5-HT1A mediated serotonergic signaling are increased.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Muguruza et al.
Neuropharmacology, 86, 311-318 (2014-08-26)
Several studies have demonstrated alterations in serotonin 5-HT2A (5-HT2AR) and glutamate metabotropic mGlu2 (mGlu2R) receptors in depression, but never in the same sample population. Recently it has been shown that both receptors form a functional receptor heterocomplex that is altered
Hun Soo Chang et al.
Journal of clinical psychopharmacology, 34(4), 446-454 (2014-06-10)
We tested for the association of HTR1A and 5-HTT genetic polymorphisms with treatment response to mirtazapine and evaluated the interactive effect between the polymorphisms in 283 patients with major depressive disorder. Korean subjects with diagnosis of major depressive disorder using
Katarzyna Kamińska et al.
Pharmacological reports : PR, 66(6), 984-990 (2014-12-03)
The aim of our study was to understand the mechanism of clinical efficacy of the combination of an antidepressant and risperidone in drug-resistant depression. We studied the effect of an antidepressant (mirtazapine) and risperidone (atypical antipsychotic), given separately or jointly
Hun Soo Chang et al.
Journal of neural transmission (Vienna, Austria : 1996), 122(1), 59-68 (2014-12-03)
Hyperactivity of the hypothalamic-pituitary-adrenal (HPA) axis is among the most consistent neuroendocrine abnormalities in major depressive disorder (MDD). The peptide adrenocorticotropin hormone (ACTH) mediates HPA axis function during stress and is encoded by the proopiomelanocortin (POMC) gene polycistronically. After screening
Michael Paulzen et al.
Psychopharmacology, 232(4), 807-813 (2014-08-26)
The aim of this study was to investigate the distribution pattern of mirtazapine and its metabolite normirtazapine (N-desmethylmirtazapine) in blood and cerebrospinal fluid (CSF). Concentrations of mirtazapine were measured in blood serum and CSF of 16 patients treated with daily

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service