Skip to Content
Merck
All Photos(1)

Key Documents

Y0000656

Yohimbine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

17-Hydroxyyohimban-16-carboxylic acid methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O3 · HCl
CAS Number:
Molecular Weight:
390.90
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

yohimbine

manufacturer/tradename

EDQM

mp

288-290 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.COC(=O)[C@H]1[C@@H](O)CCC2CN3CCc4c([nH]c5ccccc45)C3CC12

InChI

1S/C21H26N2O3.ClH/c1-26-21(25)19-15-10-17-20-14(13-4-2-3-5-16(13)22-20)8-9-23(17)11-12(15)6-7-18(19)24;/h2-5,12,15,17-19,22,24H,6-11H2,1H3;1H/t12-,15-,17-,18-,19+;/m0./s1

InChI key

PIPZGJSEDRMUAW-VJDCAHTMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Yohimbine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdellaziz Souli et al.
Journal of medicinal food, 17(7), 782-786 (2014-03-13)
The current study was performed to measure the chemical composition and the effects of dates pulp extract and palm sap on gastrointestinal transit (GIT) activity in healthy adult rats. In this respect, male Wistar rats fasted for 24 hours were
Leila Moezi et al.
Pharmacology, biochemistry, and behavior, 126, 36-42 (2014-09-23)
Adenosine has anticonvulsant effects in various models of seizures. Alpha-2 adrenoceptors have also demonstrated different effects in different models of epilepsy. In this study, the role of alpha-2 adrenoceptors in the anticonvulsant effects of adenosine in mice was determined according
Fatih İlkaya et al.
Pharmacology, biochemistry, and behavior, 122, 1-6 (2014-03-04)
It has been shown that imipramine, a tricyclic antidepressant (TCA), is a potent analgesic agent. However, the effect of imipramine on visceral pain has not been extensively investigated. In the current study, our aim was to characterise the putative analgesic
Bin Mei et al.
Brain, behavior, and immunity, 91, 296-314 (2020-10-12)
Sepsis-associated encephalopathy (SAE) is a significant clinical issue that is associated with increased mortality and cost of health care. Dexmedetomidine, an α2 adrenoceptor agonist that is used to provide sedation, has been shown to induce neuroprotection under various conditions. This
Roberto Ciccocioppo et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(11), 2601-2610 (2014-05-28)
Since its discovery, the nociceptin/orphanin FQ (N/OFQ)-NOP receptor system has been extensively investigated as a promising target to treat alcoholism. Encouraging results obtained with the endogenous ligand N/OFQ stimulated research towards the development of novel brain-penetrating NOP receptor agonists with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service