Skip to Content
Merck
All Photos(1)

Key Documents

Y0000501

Trandolapril

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(2S,3aR,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylic acid, Mavik

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34N2O5
CAS Number:
Molecular Weight:
430.54
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

trandolapril

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2C3CCCCC3C[C@H]2C(O)=O

InChI

1S/C24H34N2O5/c1-3-31-24(30)19(14-13-17-9-5-4-6-10-17)25-16(2)22(27)26-20-12-8-7-11-18(20)15-21(26)23(28)29/h4-6,9-10,16,18-21,25H,3,7-8,11-15H2,1-2H3,(H,28,29)/t16-,18+,19-,20-,21-/m0/s1

InChI key

VXFJYXUZANRPDJ-WTNASJBWSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trandolapril EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Trandolapril is an ACE inhibitor.
Trandolapril is an ACE inhibitor. Trandolapril differs from other ACE inhibitors in that it has a longer biological half-life and a high degree of lipophilicity.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Miyazaki et al.
American journal of hypertension, 8(10 Pt 2), 63S-67S (1995-10-01)
This article discusses the pharmacologic basis on which trandolapril exhibits a more potent and longer-lasting antihypertensive effect than its chemical prototype enalapril. Our studies have shown that 1) trandolapril and its active metabolite trandolaprilat are more lipophilic than enalapril and
M Buksa
Medicinski arhiv, 54(2), 103-106 (2000-08-10)
Left ventricle systolic dysfunction and congestive heart failure after AMI are still a great problem in cardiology. Introduction of angiotensin converting enzyme (ACE) inhibitors after AMI has been a new step in therapy of the left ventricle systolic disfunction. Some
C A Gaillard et al.
American heart journal, 125(5 Pt 2), 1542-1546 (1993-05-01)
In this article the clinical experiences with the new non-sulfhydryl angiotensin-converting enzyme inhibitor trandolapril are reviewed. The drug lowers blood pressure effectively in patients with mild-to-severe hypertension including elderly and obese patients. Distinguishing features of the compound are its long
J F Hansen et al.
Journal of hypertension. Supplement : official journal of the International Society of Hypertension, 15(2), S119-S122 (1997-03-01)
EFFECTS OF VERAPAMIL AND TRANDOLAPRIL: Progression of heart failure, sudden death and death from re-infarction are the major cause of the increased mortality in postinfarct patients with congestive heart failure. Angiotensin converting enzyme (ACE) inhibitors such as trandolapril can prevent
Neil A Reynolds et al.
Drugs, 65(13), 1893-1914 (2005-08-24)
Trandolapril/verapamil sustained release (SR) [Tarka] is an oral, fixed-dose combination of the ACE inhibitor trandolapril and the SR formulation of the phenylalkylamine calcium channel antagonist verapamil. It is indicated for the treatment of hypertension in patients who require more than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service