Skip to Content
Merck
All Photos(1)

Key Documents

Y0000440

Didanosine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2′,3′-Dideoxyinosine, ddI, ddIno

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4O3
CAS Number:
Molecular Weight:
236.23
Beilstein:
3619529
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

didanosine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@@H]1CC[C@@H](O1)n2cnc3C(=O)NC=Nc23

InChI

1S/C10H12N4O3/c15-3-6-1-2-7(17-6)14-5-13-8-9(14)11-4-12-10(8)16/h4-7,15H,1-3H2,(H,11,12,16)/t6-,7+/m0/s1

InChI key

BXZVVICBKDXVGW-NKWVEPMBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Didanosine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Perry et al.
Drugs, 52(6), 928-962 (1996-12-01)
Didanosine is a dideoxynucleoside analogue, which is phosphorylated to the active metabolite dideoxyadenosine triphosphate (ddATP) intracellularly. At therapeutic concentrations, ddATP inhibits HIV replication by inhibiting HIV reverse transcriptase. Didanosine is established as a first-line treatment for patients with HIV disease
M Youle
Antiviral therapy, 3 Suppl 4, 35-37 (2000-03-21)
The current focus on simplifying treatment regimens for patients with human immunodeficiency virus (HIV) infection has contributed to the interest in once-daily therapy. The triphosphate of didanosine (2',3'-dideoxyinosine or DDI) has a long intracellular half-life, which supports the use of
Masahiko Okiyama et al.
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 120(2), 115-122 (2002-08-22)
An active metabolite, ddATP, of didanosine that is an analogue of purine-nucleoside (a component of nucleic acid) was known to inhibit the activity of DNA polymerase for E. coli. In 1985, Dr. Michiya et al. of NCI reported that didanosine
Patricia Pecora Fulco et al.
The Annals of pharmacotherapy, 37(9), 1325-1328 (2003-08-19)
To evaluate the pharmacokinetic interaction between tenofovir and didanosine when used in combination as a highly active antiretroviral therapy regimen. Literature retrieval was accessed through MEDLINE (1966-January 2003) using the terms tenofovir and didanosine. Abstracts from recent meetings, including the
M Masiá et al.
International journal of STD & AIDS, 16(9), 646-648 (2005-09-24)
The combination of tenofovir and didanosine results in an increase in the didanosine plasma exposure and might augment the risk for didanosine toxicity. Although pharmacokinetic studies support a didanosine dose reduction to 250 mg when used concurrently with tenofovir in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service