Y0000394
Dipivefrine for system suitability
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Dipivefrin hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
dipivefrine
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
InChI
1S/C19H29NO5.ClH/c1-18(2,3)16(22)24-14-9-8-12(13(21)11-20-7)10-15(14)25-17(23)19(4,5)6;/h8-10,13,20-21H,11H2,1-7H3;1H
InChI key
VKFAUCPBMAGVRG-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Dipivefrine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.
Packaging
The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.
Other Notes
Sales restrictions may apply.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Contact dermatitis from dipivefrine with possible cross-reaction to epinephrine.
Contact dermatitis, 39(6), 325-326 (1999-01-05)
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 19(3), 205-215 (2003-06-28)
Over releasing of glutamate and cellular calcium influx always results in neuronal death. In the present study, we investigated various commercial antiglaucoma drugs including timolol (0.58 microM to 58 microM), betaxolol (1.62 microM to 162 microM), carteolol (6.8 microM to
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 237(2), 117-124 (1999-02-13)
A study was carried out to ascertain, in ophthalmologically normal subjects, the short-term effects of dipivefrin hydrochloride 0.1% on visual performance and make comparisons with pilocarpine. Twelve normal volunteers aged 20-26 years attended on three occasions. One eye, randomly selected
Yan ke xue bao = Eye science, 15(2), 97-102 (2003-02-13)
To establish the method of culturing calf trabecular cells (CTCs) in vitro, to understand the morphology and function of CTCs, to probe into the effect of resistance of aqueous outflow in the pathogenesis and mechanism about primary open angle glaucoma(POAG).
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society, 12(4), 217-219 (2001-12-26)
A review of the literature has identified 10 agents causing contact dermatitis among topically administered drugs for glaucoma. These agents include beta-blockers (timolol, befunolol, betaxolol, levobunolol, carteolol, metipranolol), a carbonic anhydrase inhibitor (dorzolamide), a parasympathomimetic (pilocarpine), and sympathomimetics (dipivefrin, apraclonidine).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service