Skip to Content
Merck
All Photos(1)

Key Documents

Y0000317

Mesna impurity D

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2,2′-Dithiobisethanesulfonic acid, 2,2′-(Disulphanediyl)bis(ethanesulphonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10O6S4
CAS Number:
Molecular Weight:
282.38
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mesna

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])CCSSCC[S](=O)(=O)[O-]

InChI

1S/C4H10O6S4.2Na/c5-13(6,7)3-1-11-12-2-4-14(8,9)10;;/h1-4H2,(H,5,6,7)(H,8,9,10);;/q;2*+1/p-2

InChI key

KQYGMURBTJPBPQ-UHFFFAOYSA-L

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mesna impurity D EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miranda Verschraagen et al.
Biochemical pharmacology, 68(3), 493-502 (2004-07-10)
Disodium 2,2'-dithio-bis-ethane sulfonate (BNP7787) is under investigation as a potential new chemoprotector against cisplatin-induced nephrotoxicity. The selective protection of BNP7787 appears to arise from the preferential uptake of the drug in the kidneys, where BNP7787 would undergo intracellular conversion into
E Boven et al.
British journal of cancer, 92(9), 1636-1643 (2005-04-21)
BNP7787 (disodium 2,2'-dithio-bis-ethane sulphonate; Tavocept) is a novel agent developed to protect against cisplatin (cis-diammine-dichloroplatinum(II))-associated chronic toxicities. In this study, we determined the recommended dose of BNP7787 when preceding a fixed dose of cisplatin, the pharmacokinetics (PKs) and the possible
R Todd Reilly
IDrugs : the investigational drugs journal, 7(1), 64-69 (2004-01-20)
BioNumerik Pharmacology Inc, Baxter Oncology GmbH (formerly ASTA Medica AG) and Grelan Pharmaceutical Co Ltd are developing BNP-7787 for the potential reduction of toxicity associated with cisplatin and carboplatin treatment in cancer patients.
Frederick H Hausheer et al.
Cancer chemotherapy and pharmacology, 52 Suppl 1, S3-15 (2003-06-24)
Any approach applied to drug discovery and development by the medical community and pharmaceutical industry has a direct impact on the future availability of improved, novel, and curative therapies for patients with cancer. By definition, drug discovery is a complex
Miranda Verschraagen et al.
Clinical pharmacology and therapeutics, 74(2), 157-169 (2003-08-02)
BNP7787 (disodium 2,2'-dithio-bis-ethane sulfonate) is currently undergoing development as a chemoprotective agent to prevent common and serious cisplatin-induced side effects. In the kidneys, intestine, and liver, BNP7787 is believed to undergo intracellular conversion into 2-mercaptoethane sulfonate (mesna), which can locally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service