Skip to Content
Merck
All Photos(1)

Key Documents

Y0000226

Moxonidine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methyl-5-pyrimidinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12ClN5O
CAS Number:
Molecular Weight:
241.68
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

moxonidine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1nc(nc(c1N=C2NCCN2)OC)C

InChI

1S/C9H12ClN5O/c1-5-13-7(10)6(8(14-5)16-2)15-9-11-3-4-12-9/h3-4H2,1-2H3,(H2,11,12,15)

InChI key

WPNJAUFVNXKLIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Moxonidine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scintigraphic diagnosis of Erdheim-Chester disease.
Hans Balink et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 29(16), e470-e472 (2011-03-23)
P Ernsberger
Journal of cardiovascular pharmacology, 35(7 Suppl 4), S27-S41 (2001-05-11)
The I1-imidazoline receptor is a novel neurotransmitter receptor found mainly in the brainstem, adrenal medulla and kidney. The actions of moxonidine are described at the level of individual biomolecules, cells, tissues, organs and finally with integrative functions. The receptor functions
B N Prichard et al.
Journal of hypertension. Supplement : official journal of the International Society of Hypertension, 15(1), S47-S55 (1997-01-01)
Imidazoline I1-receptor agonism represents a new mode of antihypertensive action to inhibit peripheral alpha-adrenergic tone by a central mechanism. Adrenaline, noradrenaline and renin levels are reduced, a finding consistent with central inhibition of sympathetic tone. Acute haemodynamic studies indicate that
Angelita Maria Stabile et al.
Life sciences, 88(11-12), 493-503 (2011-02-01)
Heart failure is a progressive deterioration in heart function associated with overactivity of the sympathetic nervous system. The benefit of inhibition of sympathetic activity by moxonidine, a centrally acting imidazoline receptor agonist, was questioned based on the outcome of a
P A van Zwieten
Journal of hypertension, 15(2), 117-125 (1997-02-01)
Clonidine, guanfacine, guanabenz and alpha-methyl-dioxyphenylalanine (DOPA), the prototypes of centrally acting antihypertensives, are assumed to induce peripheral sympathoinhibition and a reduction in blood pressure via the stimulation of alpha2-adrenoceptors in the brain stem. More recently, central imidazoline (I1)-receptors have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service