Skip to Content
Merck
All Photos(1)

Key Documents

Y0000041

Josamycin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H69NO15
CAS Number:
Molecular Weight:
827.99
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

josamycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CO[C@H]1[C@@H](CC(=O)O[C@H](C)C\C=C\C=C\[C@H](O)[C@H](C)C[C@H](CC=O)[C@@H]1O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](OC(=O)CC(C)C)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)OC(C)=O

InChI

1S/C42H69NO15/c1-23(2)19-32(47)56-40-27(6)53-34(22-42(40,8)50)57-37-26(5)54-41(36(49)35(37)43(9)10)58-38-29(17-18-44)20-24(3)30(46)16-14-12-13-15-25(4)52-33(48)21-31(39(38)51-11)55-28(7)45/h12-14,16,18,23-27,29-31,34-41,46,49-50H,15,17,19-22H2,1-11H3/b13-12+,16-14+/t24-,25-,26-,27+,29+,30+,31-,34+,35-,36-,37-,38+,39+,40+,41+,42-/m1/s1

InChI key

XJSFLOJWULLJQS-NGVXBBESSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Josamycin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Przybylski et al.
Magnetic resonance in chemistry : MRC, 48(4), 286-296 (2010-02-27)
Four new hydroxy-aminoalkyl derivatives of alpha,beta-unsaturated macrolide-josamycin (2-5) have been synthesised and their structures have been studied by means of (1)H and (13)C NMR and FT-IR methods. Complete assignment of resonances in the (1)H and (13)C NMR spectra has been
M Weidlich et al.
Letters in applied microbiology, 46(2), 155-159 (2007-11-22)
Evaluation of the activity profiles of standard prokaryotic translation inhibitors with different physicochemical properties under whole-cell and cell-free conditions. The minimal inhibitory concentration values (cell-free/whole-cell microg ml(-1)) for three aminoglycosides (neomycin, 0.01/6.92; paromomycin, 0.7/1.96; streptomycin 1.45/1.57), three macrolides (erythromycin, 1.53/56.9;
Yu-Hong Min et al.
Archives of pharmacal research, 31(3), 377-380 (2008-04-15)
Frequencies of spontaneous mutation from inducible resistance to constitutive resistance were determined for the four clinical isolates of erythromycin-resistant enterococci, including one isolate with ermB gene and three clinical isolates with ermA gene. The rate of ermB mutation was higher
Martin Lovmar et al.
Biochimie, 91(8), 989-995 (2009-05-26)
Macrolide antibiotics block the entrance of nascent peptides to the peptide exit tunnel of the large ribosomal subunit. Expression of specific cis-acting peptides confers low-level macrolide-resistance. We show that, in the case of josamycin, peptide expression does not eject josamycin
Takeshi Shimizu et al.
Mediators of inflammation, 2012, 265714-265714 (2012-05-12)
To examine the in vivo effects of the 15-member macrolide, azithromycin (AZM), on mucus hypersecretion, we induced hypertrophic and metaplastic changes of goblet cells in rat nasal epithelium by intranasal instillation of ovalbumin (OVA) in OVA-sensitized rats, or by intranasal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service