Skip to Content
Merck
All Photos(1)

Key Documents

V0400000

Vincristine sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Vincristine sulfate salt, 22-Oxovincaleukoblastine sulfate salt, Leurocristine sulfate salt, VCR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H56N4O10 · H2SO4
CAS Number:
Molecular Weight:
923.04
Beilstein:
3924631
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

vincristine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−70°C

SMILES string

OS(O)(=O)=O.CC[C@]1(O)CC2CN(CCc3c([nH]c4ccccc34)[C@@](C2)(C(=O)OC)c5cc6c(cc5OC)N(C=O)C7[C@](O)([C@H](OC(C)=O)[C@]8(CC)C=CCN9CC[C@]67C89)C(=O)OC)C1

InChI

1S/C46H56N4O10.H2O4S/c1-7-42(55)22-28-23-45(40(53)58-5,36-30(14-18-48(24-28)25-42)29-12-9-10-13-33(29)47-36)32-20-31-34(21-35(32)57-4)50(26-51)38-44(31)16-19-49-17-11-15-43(8-2,37(44)49)39(60-27(3)52)46(38,56)41(54)59-6;1-5(2,3)4/h9-13,15,20-21,26,28,37-39,47,55-56H,7-8,14,16-19,22-25H2,1-6H3;(H2,1,2,3,4)/t28-,37-,38+,39+,42-,43+,44+,45-,46-;/m0./s1

InChI key

AQTQHPDCURKLKT-PNYVAJAMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Vincristine sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Muta. 2 - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shipra Malik et al.
Cell reports. Medicine, 5(1), 101354-101354 (2024-01-07)
Targeting oncogenes at the genomic DNA level can open new avenues for precision medicine. Significant efforts are ongoing to target oncogenes using RNA-targeted and protein-targeted platforms, but no progress has been made to target genomic DNA for cancer therapy. Here
Fei San Lee et al.
PloS one, 19(5), e0300254-e0300254 (2024-05-02)
Low back pain, knee osteoarthritis, and cancer patients suffer from chronic pain. Aberrant nerve growth into intervertebral disc, knee, and tumors, are common pathologies that lead to these chronic pain conditions. Axonal dieback induced by capsaicin (Caps) denervation has been
Retraction notice to " IP1867B suppresses the Insulin-like Growth Factor 1 Receptor (IGF1R) ablating epidermal growth factor receptor inhibitor resistance in adult high grade gliomas" [Canc. Lett., 458 (2019) pages 29-38].
K Mihajluk et al.
Cancer letters, 507, 39-39 (2021-03-21)
Hanbing Song et al.
Nature communications, 13(1), 4878-4878 (2022-08-26)
Pediatric hepatoblastoma is the most common primary liver cancer in infants and children. Studies of hepatoblastoma that focus exclusively on tumor cells demonstrate sparse somatic mutations and a common cell of origin, the hepatoblast, across patients. In contrast to the
K Mihajluk et al.
Cancer letters, 458, 29-38 (2019-05-28)
This article has been retracted at the request of the Editor-in-Chief due to concerns regarding the legitimacy of images and data presented in the paper. Though a corrigendum (Can. Lett. Vol. 469, 2020, pages 524-535) was previously published to address

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service